Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00710297_m1
          See other CYP27A1 GE Assays ›
          Gene Symbol:
          Cyp27a1
          Gene Name:
          cytochrome P450, family 27, subfamily a, polypeptide 1
          Gene Aliases:
          Cyp27, P450C27
          Chromosome Location:
          Chr.9: 81968289 - 81998213 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00710297_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp27a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC061848.1 - 1 - 2 301 5621918 63
          M38566.1 - 1 - 2 302 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900000908.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          oxygenase

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P15847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935874 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152991 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99882 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3984 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101615 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15847 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15847 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P15847 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15847 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19800 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20014 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10924417 - -
          Affymetrix Rat Gene 1.0 ST Array 10924420 - -
          Affymetrix Rat Gene 1.0 ST Array 10924413 - -
          Affymetrix Rat Gene 1.0 ST Array 10924415 - -
          Affymetrix Rat Gene 1.0 ST Array 10924421 - -
          Affymetrix Rat Gene 1.0 ST Array 10924414 - -
          Affymetrix Rat Gene 1.0 ST Array 10924416 - -
          Affymetrix Rat Gene 1.0 ST Array 10924418 - -
          Affymetrix Rat Gene 1.0 ST Array 10924419 - -
          Affymetrix Rat Gene 1.1 ST Array 10924411 - -
          Rat Genome 230 2.0 Array 1387914_at - -
          Rat Genome 230 2.0 Array 1387914_PM_at - -
          Affymetrix HT Rat Focus Array 1387914_at - -
          Affymetrix HT Rat Focus Array 1387914_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0900007592.rn.1 TC0900000908.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900007591.rn.1 TC0900000908.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900007594.rn.1 TC0900000908.rn.1 RTA TC
          Rat Expression Set 230 1387914_at - -
          Rat Expression Set 230 1387914_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10924411 - -
          Rat Genome U34 Set Y07534cds_s_at - -
          Rat Genome U34 Set M38566mRNA_s_at - -
          Rat Genome U34 Set Y07534cds_s_at - -
          Rat Genome U34 Set M38566mRNA_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22167804 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376833 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lq2bv:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0