Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00710355_m1
          See other ARFGEF2 GE Assays ›
          Gene Symbol:
          Arfgef2
          Gene Name:
          ADP-ribosylation factor guanine nucleotide-exchange factor 2 (brefeldin A-inhibited)
          Gene Aliases:
          Big2
          Chromosome Location:
          Chr.3: 163570437 - 163656612 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00710355_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arfgef2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_181083.2 NP_851597.2 33 - 34 4532 78
          XM_006235640.2 XP_006235702.1 33 - 34 4714 78
          GenBank mRNA AY255526.2 - 33 - 34 4532 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002351.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Arfgef2

          Panther Classification:

          Molecular Function -

          guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          receptor recycling
          exocytosis
          Golgi to plasma membrane transport
          endosome organization
          endomembrane system organization
          protein transport
          regulation of ARF protein signal transduction
          positive regulation of tumor necrosis factor production
          intracellular signal transduction
          positive regulation of GTPase activity
          guanyl-nucleotide exchange factor activity
          ARF guanyl-nucleotide exchange factor activity
          protein binding
          myosin binding
          protein kinase A regulatory subunit binding
          GABA receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1012529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737047 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71238 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95100 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49682 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89092 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6753 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69998 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30720 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13004 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69596 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65514 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1012529 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1012529 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1012529 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1012529 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P21444 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30325 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13621 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71523 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21915 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92129 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10842354 - -
          Affymetrix Rat Gene 1.0 ST Array 10842359 - -
          Affymetrix Rat Gene 1.0 ST Array 10842361 - -
          Affymetrix Rat Gene 1.0 ST Array 10842371 - -
          Affymetrix Rat Gene 1.0 ST Array 10842351 - -
          Affymetrix Rat Gene 1.0 ST Array 10842357 - -
          Affymetrix Rat Gene 1.0 ST Array 10842365 - -
          Affymetrix Rat Gene 1.0 ST Array 10842378 - -
          Affymetrix Rat Gene 1.0 ST Array 10842362 - -
          Affymetrix Rat Gene 1.0 ST Array 10842379 - -
          Affymetrix Rat Gene 1.0 ST Array 10842360 - -
          Affymetrix Rat Gene 1.0 ST Array 10842380 - -
          Affymetrix Rat Gene 1.0 ST Array 10842364 - -
          Affymetrix Rat Gene 1.0 ST Array 10842370 - -
          Affymetrix Rat Gene 1.0 ST Array 10842356 - -
          Affymetrix Rat Gene 1.0 ST Array 10842353 - -
          Affymetrix Rat Gene 1.0 ST Array 10842347 - -
          Affymetrix Rat Gene 1.0 ST Array 10842348 - -
          Affymetrix Rat Gene 1.0 ST Array 10842381 - -
          Affymetrix Rat Gene 1.0 ST Array 10842358 - -
          Affymetrix Rat Gene 1.0 ST Array 10842369 - -
          Affymetrix Rat Gene 1.0 ST Array 10842377 - -
          Affymetrix Rat Gene 1.0 ST Array 10842382 - -
          Affymetrix Rat Gene 1.0 ST Array 10842345 - -
          Affymetrix Rat Gene 1.0 ST Array 10842350 - -
          Affymetrix Rat Gene 1.0 ST Array 10842373 - -
          Affymetrix Rat Gene 1.0 ST Array 10842355 - -
          Affymetrix Rat Gene 1.0 ST Array 10842367 - -
          Affymetrix Rat Gene 1.0 ST Array 10842366 - -
          Affymetrix Rat Gene 1.0 ST Array 10842349 - -
          Affymetrix Rat Gene 1.0 ST Array 10842346 - -
          Affymetrix Rat Gene 1.0 ST Array 10842374 - -
          Affymetrix Rat Gene 1.0 ST Array 10842375 - -
          Affymetrix Rat Gene 1.0 ST Array 10842368 - -
          Affymetrix Rat Gene 1.0 ST Array 10842363 - -
          Affymetrix Rat Gene 1.0 ST Array 10842376 - -
          Affymetrix Rat Gene 1.0 ST Array 10842372 - -
          Affymetrix Rat Gene 1.0 ST Array 10842352 - -
          Affymetrix Rat Gene 1.0 ST Array 10842383 - -
          Affymetrix Rat Gene 1.1 ST Array 10842344 - -
          Rat Genome 230 2.0 Array 1374213_PM_at - -
          Rat Genome 230 2.0 Array 1374213_at - -
          Affymetrix HT Rat Focus Array 1374213_PM_at - -
          Affymetrix HT Rat Focus Array 1374213_at - -
          Affymetrix Rat Clariom S Assay PSR0300017551.rn.1 TC0300002351.rn.1 RTA TC
          Rat Expression Set 230 1374213_at - -
          Rat Expression Set 230 1374213_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10842344 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21238108 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366511 - -
          RatRef-12 Expression BeadChip ILMN_59859 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0