Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00710376_m1
          See other CYP26B1 GE Assays ›
          Gene Symbol:
          Cyp26b1
          Gene Name:
          cytochrome P450, family 26, subfamily b, polypeptide 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 116261796 - 116278615 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00710376_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp26b1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_181087.2 NP_851601.1 2 - 3 588 64
          GenBank mRNA AY245532.2 - 2 - 3 588 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400003200.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyp26b1

          Panther Classification:

          Molecular Function -

          oxygenase

          Gene Ontology Categories:

          Function(s) Process(es)

          cell fate determination
          establishment of T cell polarity
          inflammatory response
          male meiosis
          spermatogenesis
          proximal/distal pattern formation
          positive regulation of gene expression
          embryonic limb morphogenesis
          retinoic acid catabolic process
          retinoic acid metabolic process
          tongue morphogenesis
          regulation of T cell differentiation
          retinoic acid receptor signaling pathway
          regulation of retinoic acid receptor signaling pathway
          negative regulation of retinoic acid receptor signaling pathway
          oxidation-reduction process
          bone morphogenesis
          establishment of skin barrier
          cornification
          cellular response to retinoic acid
          positive regulation of tongue muscle cell differentiation
          retinoic acid binding
          iron ion binding
          retinoic acid 4-hydroxylase activity
          heme binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P693964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383184 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46776 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70067 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83798 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P693964 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P693964 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P693964 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P693964 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74361 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55742 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10863611 - -
          Affymetrix Rat Gene 1.0 ST Array 10863614 - -
          Affymetrix Rat Gene 1.0 ST Array 10863610 - -
          Affymetrix Rat Gene 1.0 ST Array 10863609 - -
          Affymetrix Rat Gene 1.0 ST Array 10863613 - -
          Affymetrix Rat Gene 1.0 ST Array 10863612 - -
          Affymetrix Rat Gene 1.1 ST Array 10863608 - -
          Rat Genome 230 2.0 Array 1376667_PM_at - -
          Rat Genome 230 2.0 Array 1384392_at - -
          Rat Genome 230 2.0 Array 1376667_at - -
          Rat Genome 230 2.0 Array 1384392_PM_at - -
          Affymetrix HT Rat Focus Array 1384392_PM_at - -
          Affymetrix HT Rat Focus Array 1376667_PM_at - -
          Affymetrix HT Rat Focus Array 1384392_at - -
          Affymetrix HT Rat Focus Array 1376667_at - -
          Affymetrix Rat Clariom S Assay PSR0400022737.rn.1 TC0400003200.rn.1 RTA TC
          Rat Expression Set 230 1376667_PM_at - -
          Rat Expression Set 230 1376667_at - -
          Rat Expression Set 230 1384392_at - -
          Rat Expression Set 230 1384392_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10863608 - -
          Rat Genome U34 Set rc_AA892066_at - -
          Rat Genome U34 Set rc_AA957564_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20744053 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363580 - -
          RatRef-12 Expression BeadChip ILMN_64141 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline