Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00710421_m1
          See other SLC38A2 GE Assays ›
          Gene Symbol:
          Slc38a2
          Gene Name:
          solute carrier family 38, member 2
          Gene Aliases:
          Ata2, Atrc2, Sat2, Snat2
          Chromosome Location:
          Chr.7: 138088654 - 138100869 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00710421_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc38a2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_181090.2 NP_851604.1 15 - 16 1808 57
          XM_006242282.1 XP_006242344.1 14 - 15 1941 57
          XM_006242283.1 XP_006242345.1 13 - 14 1825 57
          GenBank mRNA AF173682.2 - 15 - 16 1609 57
          AF249673.1 - 15 - 16 1808 57
          AF273024.1 - 15 - 16 1643 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700003804.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Transporter 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc38a2

          Panther Classification:

          Molecular Function -

          amino acid transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          amino acid transmembrane transport
          sodium ion transport
          amino acid transport
          amino acid transmembrane transporter activity
          symporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P104652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243301 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23141 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18093 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P104652 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P104652 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P104652 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P463878 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P104652 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11008 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76758 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10906615 - -
          Affymetrix Rat Gene 1.0 ST Array 10906610 - -
          Affymetrix Rat Gene 1.0 ST Array 10906612 - -
          Affymetrix Rat Gene 1.0 ST Array 10906617 - -
          Affymetrix Rat Gene 1.0 ST Array 10906621 - -
          Affymetrix Rat Gene 1.0 ST Array 10906611 - -
          Affymetrix Rat Gene 1.0 ST Array 10906614 - -
          Affymetrix Rat Gene 1.0 ST Array 10906609 - -
          Affymetrix Rat Gene 1.0 ST Array 10906619 - -
          Affymetrix Rat Gene 1.0 ST Array 10906623 - -
          Affymetrix Rat Gene 1.0 ST Array 10906625 - -
          Affymetrix Rat Gene 1.0 ST Array 10906616 - -
          Affymetrix Rat Gene 1.0 ST Array 10906613 - -
          Affymetrix Rat Gene 1.0 ST Array 10906624 - -
          Affymetrix Rat Gene 1.0 ST Array 10906622 - -
          Affymetrix Rat Gene 1.0 ST Array 10906620 - -
          Affymetrix Rat Gene 1.0 ST Array 10906618 - -
          Affymetrix Rat Gene 1.1 ST Array 10906608 - -
          Affymetrix Rat Clariom S Assay PSR0700030304.rn.1 TC0700003804.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700030305.rn.1 TC0700003804.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700030307.rn.1 TC0700003804.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10906608 - -
          Rat Genome U34 Set rc_AA963102_at - -
          Rat Genome U34 Set rc_AI171231_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21428024 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359139 - -
          RatRef-12 Expression BeadChip ILMN_47981 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline