Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00710561_m1
          See other PPP2R5B GE Assays ›
          Gene Symbol:
          Ppp2r5b
          Gene Name:
          protein phosphatase 2, regulatory subunit B', beta
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 221597294 - 221605467 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00710561_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppp2r5b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_181379.2 NP_852044.1 10 - 11 1160 82
          XM_008760143.1 XP_008758365.1 11 - 12 1731 82
          GenBank mRNA AY251278.1 - 10 - 11 1113 82
          BC090324.1 - 10 - 11 1160 7128996 82

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ppp2r5b

          Panther Classification:

          Molecular Function -

          protein phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of protein phosphorylation
          signal transduction
          regulation of receptor activity
          positive regulation of neuron projection development
          positive regulation of protein complex assembly
          regulation of protein autophosphorylation
          positive regulation of transcription from RNA polymerase II promoter
          regulation of peptidyl-tyrosine phosphorylation
          regulation of catalytic activity
          positive regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of neurotrophin TRK receptor signaling pathway
          negative regulation of G0 to G1 transition
          positive regulation of cell cycle arrest
          cellular response to growth factor stimulus
          protein phosphatase type 2A regulator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P463480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141617 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85704 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4955 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P463480 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P463480 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P463480 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P463480 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1180 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16144 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10728165 - -
          Affymetrix Rat Gene 1.0 ST Array 10728168 - -
          Affymetrix Rat Gene 1.0 ST Array 10728178 - -
          Affymetrix Rat Gene 1.0 ST Array 10728167 - -
          Affymetrix Rat Gene 1.0 ST Array 10728169 - -
          Affymetrix Rat Gene 1.0 ST Array 10728174 - -
          Affymetrix Rat Gene 1.0 ST Array 10728172 - -
          Affymetrix Rat Gene 1.0 ST Array 10728171 - -
          Affymetrix Rat Gene 1.0 ST Array 10728175 - -
          Affymetrix Rat Gene 1.0 ST Array 10728170 - -
          Affymetrix Rat Gene 1.0 ST Array 10728166 - -
          Affymetrix Rat Gene 1.0 ST Array 10728177 - -
          Affymetrix Rat Gene 1.0 ST Array 10728173 - -
          Affymetrix Rat Gene 1.0 ST Array 10728176 - -
          Affymetrix Rat Gene 1.1 ST Array 10728164 - -
          Rat Genome 230 2.0 Array 1377235_a_at - -
          Rat Genome 230 2.0 Array 1377235_PM_a_at - -
          Affymetrix HT Rat Focus Array 1377235_PM_a_at - -
          Affymetrix HT Rat Focus Array 1377235_a_at - -
          Affymetrix Rat Clariom S Assay PSR0100057483.rn.1 TC0100007162.rn.1 RTA TC
          Rat Expression Set 230 1377235_a_at - -
          Rat Expression Set 230 1377235_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10728164 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21826659 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1649757 - -
          RatRef-12 Expression BeadChip ILMN_57177 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline