Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn00710603_m1
See other TNFSF9 GE Assays ›
Gene Symbol:
Tnfsf9
Gene Name:
tumor necrosis factor (ligand) superfamily, member 9
Gene Aliases:
-
Chromosome Location:
Chr.9: 9909942 - 9912276 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn00710603_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 250 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Inventoried
Catalog #
4331182
Price
284.00
Your Price
Online offer:
266.65
266.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Tnfsf9
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_181384.2
NP_852049.1
2 - 3
566
62
GenBank mRNA
AY259541.1
-
2 - 3
566
62
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0900001448.rn.1
Set Membership:
Ambion siRNA available
Amplicon length less than or equal to 70
3' Most
Recommended Assay
Rattus norvegicus
1700
Inventoried
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene Tnfsf9
Gene Ontology Categories:
Function(s)
Process(es)
immune response
positive regulation of interferon-gamma production
positive regulation of interleukin-12 production
positive regulation of interleukin-6 production
positive regulation of activated T cell proliferation
regulation of cell proliferation
myeloid dendritic cell differentiation
positive regulation of cytotoxic T cell differentiation
molecular_function
receptor binding
cytokine activity
tumor necrosis factor receptor binding
tumor necrosis factor receptor superfamily binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name
Probe IDs
Targets
Target Type
Agilent Whole Rat Genome Microarray 4x44K v2
A_44_P1004840
-
-
Agilent Whole Rat Genome Microarray 4x44K v3
A_44_P1004840
-
-
Agilent Whole Rat Genome Microarray 4x44K v3
A_44_P1004840
-
-
Agilent SurePrint G3 Rat GE 8x60K Microarray
A_44_P1004840
-
-
SurePrint G3 Rat Gene Expression Microarray G4131A
A_44_P1004840
-
-
Array Name
Probe IDs
Targets
Target Type
Affymetrix Rat Gene 1.0 ST Array
10940589
-
-
Affymetrix Rat Gene 1.1 ST Array
10940589
-
-
Rat Genome 230 2.0 Array
1377554_PM_at
-
-
Rat Genome 230 2.0 Array
1377554_at
-
-
Affymetrix HT Rat Focus Array
1377554_at
-
-
Affymetrix HT Rat Focus Array
1377554_PM_at
-
-
Affymetrix Rat Clariom S Assay
PSR0900011950.rn.1
TC0900001448.rn.1
RTA TC
Rat Expression Set 230
1377554_PM_at
-
-
Rat Expression Set 230
1377554_at
-
-
Affymetrix Rat Gene 1.0 ST Array
10940589
-
-
Array Name
Probe IDs
Targets
Target Type
ABI Rat Genome Survey Microarray
21603216
-
-
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline