Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00710611_m1
          See other LHFPL4 GE Assays ›
          Gene Symbol:
          Lhfpl4
          Gene Name:
          lipoma HMGIC fusion partner-like 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 145117533 - 145147397 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00710611_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lhfpl4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_181387.2 NP_852052.1 2 - 3 588 60
          GenBank mRNA AY278321.1 - 2 - 3 588 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400003530.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lhfpl4

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P060824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72155 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104729 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177570 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P060824 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P060824 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P060824 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P123296 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75275 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45318 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30079 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10864618 - -
          Affymetrix Rat Gene 1.0 ST Array 10864619 - -
          Affymetrix Rat Gene 1.0 ST Array 10864617 - -
          Affymetrix Rat Gene 1.0 ST Array 10864620 - -
          Affymetrix Rat Gene 1.1 ST Array 10864616 - -
          Rat Genome 230 2.0 Array 1377958_PM_at - -
          Rat Genome 230 2.0 Array 1395649_PM_at - -
          Rat Genome 230 2.0 Array 1394520_at - -
          Rat Genome 230 2.0 Array 1377958_at - -
          Rat Genome 230 2.0 Array 1394520_PM_at - -
          Rat Genome 230 2.0 Array 1395649_at - -
          Affymetrix HT Rat Focus Array 1395649_PM_at - -
          Affymetrix HT Rat Focus Array 1395649_at - -
          Affymetrix HT Rat Focus Array 1377958_at - -
          Affymetrix HT Rat Focus Array 1377958_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400024739.rn.1 TC0400003530.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400024740.rn.1 TC0400003530.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400024741.rn.1 TC0400003530.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400024738.rn.1 TC0400003530.rn.1 RTA TC
          Rat Expression Set 230 1395649_PM_at - -
          Rat Expression Set 230 1395649_at - -
          Rat Expression Set 230 1377958_at - -
          Rat Expression Set 230 1377958_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10864616 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21640207 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371076 - -
          RatRef-12 Expression BeadChip ILMN_51132 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline