Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00710638_m1
          See other NSMAF GE Assays ›
          Gene Symbol:
          Nsmaf
          Gene Name:
          neutral sphingomyelinase (N-SMase) activation associated factor
          Gene Aliases:
          FAN
          Chromosome Location:
          Chr.5: 19499111 - 19560304 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00710638_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nsmaf
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_181389.2 NP_852054.1 23 - 24 1969 135
          XM_008763544.1 XP_008761766.1 23 - 24 3028 135
          XM_008763545.1 XP_008761767.1 22 - 23 2851 135
          GenBank mRNA AY163240.1 - 23 - 24 1969 135

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500002269.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nsmaf

          Gene Ontology Categories:

          Function(s) Process(es)

          endosomal transport
          positive regulation of apoptotic process
          positive regulation of catalytic activity
          phospholipid binding
          sphingomyelin phosphodiesterase activator activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P308687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418105 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156371 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128016 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5919 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47574 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149574 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P308687 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P308687 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P308687 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P308687 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47056 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15540 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46286 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10875351 - -
          Affymetrix Rat Gene 1.0 ST Array 10875342 - -
          Affymetrix Rat Gene 1.0 ST Array 10875339 - -
          Affymetrix Rat Gene 1.0 ST Array 10875354 - -
          Affymetrix Rat Gene 1.0 ST Array 10875350 - -
          Affymetrix Rat Gene 1.0 ST Array 10875358 - -
          Affymetrix Rat Gene 1.0 ST Array 10875347 - -
          Affymetrix Rat Gene 1.0 ST Array 10875352 - -
          Affymetrix Rat Gene 1.0 ST Array 10875345 - -
          Affymetrix Rat Gene 1.0 ST Array 10875340 - -
          Affymetrix Rat Gene 1.0 ST Array 10875337 - -
          Affymetrix Rat Gene 1.0 ST Array 10875349 - -
          Affymetrix Rat Gene 1.0 ST Array 10875361 - -
          Affymetrix Rat Gene 1.0 ST Array 10875348 - -
          Affymetrix Rat Gene 1.0 ST Array 10875344 - -
          Affymetrix Rat Gene 1.0 ST Array 10875357 - -
          Affymetrix Rat Gene 1.0 ST Array 10875336 - -
          Affymetrix Rat Gene 1.0 ST Array 10875356 - -
          Affymetrix Rat Gene 1.0 ST Array 10875359 - -
          Affymetrix Rat Gene 1.0 ST Array 10875334 - -
          Affymetrix Rat Gene 1.0 ST Array 10875360 - -
          Affymetrix Rat Gene 1.0 ST Array 10875355 - -
          Affymetrix Rat Gene 1.0 ST Array 10875338 - -
          Affymetrix Rat Gene 1.0 ST Array 10875335 - -
          Affymetrix Rat Gene 1.0 ST Array 10875333 - -
          Affymetrix Rat Gene 1.0 ST Array 10875341 - -
          Affymetrix Rat Gene 1.0 ST Array 10875343 - -
          Affymetrix Rat Gene 1.0 ST Array 10875332 - -
          Affymetrix Rat Gene 1.1 ST Array 10875331 - -
          Rat Genome 230 2.0 Array 1374014_at - -
          Rat Genome 230 2.0 Array 1374014_PM_at - -
          Affymetrix HT Rat Focus Array 1374014_PM_at - -
          Affymetrix HT Rat Focus Array 1374014_at - -
          Affymetrix Rat Clariom S Assay PSR0500017197.rn.1 TC0500002269.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500017198.rn.1 TC0500002269.rn.1 RTA TC
          Rat Expression Set 230 1374014_PM_at - -
          Rat Expression Set 230 1374014_at - -
          Affymetrix Rat Gene 1.0 ST Array 10875331 - -
          Rat Genome U34 Set rc_AI229854_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22203777 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline