Search
Search
| Gene Symbol: Prss21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181477.2 | NP_852142.1 | 4 - 5 | 607 | 114 | |
| XM_006245872.2 | XP_006245934.1 | 4 - 5 | 1025 | 114 | ||
| GenBank mRNA | AB074516.1 | - | 4 - 5 | 607 | 114 | |
Set Membership: |
|