Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00710736_m1
          See other KIR3DL1 GE Assays ›
          Gene Symbol:
          Kir3dl1
          Gene Name:
          killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail, 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 73307563 - 73344955 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00710736_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kir3dl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_181479.2 NP_852144.1 5 - 6 952 77
          GenBank mRNA AF527797.1 - 5 - 6 952 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100004807.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kir3dl1

          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P140328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34429 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86630 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162509 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111527 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P140328 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P140328 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P140328 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P348802 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1607 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91414 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7971 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10718952 - -
          Affymetrix Rat Gene 1.0 ST Array 10718950 - -
          Affymetrix Rat Gene 1.0 ST Array 10718949 - -
          Affymetrix Rat Gene 1.0 ST Array 10718945 - -
          Affymetrix Rat Gene 1.0 ST Array 10718953 - -
          Affymetrix Rat Gene 1.0 ST Array 10718951 - -
          Affymetrix Rat Gene 1.0 ST Array 10718947 - -
          Affymetrix Rat Gene 1.0 ST Array 10718946 - -
          Affymetrix Rat Gene 1.1 ST Array 10718944 - -
          Affymetrix Rat Clariom S Assay PSR0100037855.rn.1 TC0100004807.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100037859.rn.1 TC0100004807.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100037854.rn.1 TC0100004807.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100037856.rn.1 TC0100004807.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100037860.rn.1 TC0100004807.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100037858.rn.1 TC0100004807.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100037857.rn.1 TC0100004807.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100037861.rn.1 TC0100004807.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10718944 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21311682 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372176 - -
          RatRef-12 Expression BeadChip ILMN_60756 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline