Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00711083_m1
          See other RNASEL GE Assays ›
          Gene Symbol:
          Rnasel
          Gene Name:
          ribonuclease L (2',5'-oligoisoadenylate synthetase-dependent)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 71188717 - 71202636 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00711083_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnasel
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_182673.2 NP_872614.1 2 - 3 1607 90
          XM_006249998.2 XP_006250060.1 4 - 5 1994 90
          XM_006249999.2 XP_006250061.1 4 - 5 2198 90
          XM_006250000.2 XP_006250062.1 4 - 5 2284 90
          XM_006250001.2 XP_006250063.1 4 - 5 1973 90
          XM_006250002.2 XP_006250064.1 3 - 4 2049 90
          XM_006250004.2 XP_006250066.1 3 - 4 2561 90
          XM_008769653.1 XP_008767875.1 3 - 4 2561 90
          GenBank mRNA AY262823.1 - 2 - 3 1609 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300000501.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rnasel

          Gene Ontology Categories:

          Function(s) Process(es)

          rRNA processing
          mRNA processing
          protein phosphorylation
          regulation of mRNA stability
          negative regulation of viral genome replication
          fat cell differentiation
          positive regulation of transcription from RNA polymerase II promoter
          defense response to virus
          RNA phosphodiester bond hydrolysis
          positive regulation of glucose import in response to insulin stimulus
          RNA binding
          ribonuclease activity
          protein kinase activity
          ATP binding
          rRNA binding
          ribonucleoprotein complex binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P160846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513601 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19086 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P160846 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P160846 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P160846 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P160846 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73149 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10764783 - -
          Affymetrix Rat Gene 1.0 ST Array 10764784 - -
          Affymetrix Rat Gene 1.0 ST Array 10764782 - -
          Affymetrix Rat Gene 1.0 ST Array 10764787 - -
          Affymetrix Rat Gene 1.0 ST Array 10764785 - -
          Affymetrix Rat Gene 1.0 ST Array 10764786 - -
          Affymetrix Rat Gene 1.1 ST Array 10764781 - -
          Rat Genome 230 2.0 Array 1380425_at - -
          Rat Genome 230 2.0 Array 1380425_PM_at - -
          Rat Genome 230 2.0 Array 1377116_at - -
          Rat Genome 230 2.0 Array 1377116_PM_at - -
          Affymetrix HT Rat Focus Array 1380425_PM_at - -
          Affymetrix HT Rat Focus Array 1377116_at - -
          Affymetrix HT Rat Focus Array 1380425_at - -
          Affymetrix HT Rat Focus Array 1377116_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300003539.rn.1 TC1300000501.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300003538.rn.1 TC1300000501.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300003537.rn.1 TC1300000501.rn.1 RTA TC
          Rat Expression Set 230 1377116_at - -
          Rat Expression Set 230 1377116_PM_at - -
          Rat Expression Set 230 1380425_PM_at - -
          Rat Expression Set 230 1380425_at - -
          Affymetrix Rat Gene 1.0 ST Array 10764781 - -
          Rat Genome U34 Set rc_AI012859_at - -
          Rat Genome U34 Set rc_AI137260_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21421685 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357295 - -
          RatRef-12 Expression BeadChip ILMN_70341 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline