Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00754905_g1
          See other CALM3 GE Assays ›
          Gene Symbol:
          Calm3
          Gene Name:
          calmodulin 3
          Gene Aliases:
          RNCAMIII
          Chromosome Location:
          Chr.1: 78844520 - 78851628 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00754905_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Calm3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012518.3 NP_036650.1 3 - 4 292 70
          GenBank mRNA BC063187.1 - 387 6890866 70
          FQ212716.1 - 358 70
          M16659.1 - 3 - 4 292 70
          X13817.1 - 4 - 5 384 70

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Calm3

          Panther Classification:

          Molecular Function -

          calmodulin-related

          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          response to amphetamine
          regulation of heart rate
          detection of calcium ion
          activation of adenylate cyclase activity
          regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
          calcium-mediated signaling
          substantia nigra development
          regulation of cytokinesis
          positive regulation of phosphoprotein phosphatase activity
          positive regulation of protein dephosphorylation
          positive regulation of nitric-oxide synthase activity
          positive regulation of cyclic-nucleotide phosphodiesterase activity
          regulation of cardiac muscle contraction
          regulation of ryanodine-sensitive calcium-release channel activity
          positive regulation of ryanodine-sensitive calcium-release channel activity
          regulation of store-operated calcium channel activity
          regulation of high voltage-gated calcium channel activity
          calcium ion binding
          protein binding
          adenylate cyclase binding
          protein kinase binding
          enzyme regulator activity
          nitric-oxide synthase regulator activity
          titin binding
          thioesterase binding
          N-terminal myristoylation domain binding
          phospholipase binding
          ion channel binding
          protein N-terminus binding
          nitric-oxide synthase binding
          protein phosphatase activator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P184465 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P019007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211549 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103557 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P184465 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P019007 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P184465 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P019007 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P019007 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P184465 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P184465 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1027005 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76819 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19720 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10719296 - -
          Affymetrix Rat Gene 1.0 ST Array 10719298 - -
          Affymetrix Rat Gene 1.0 ST Array 10719295 - -
          Affymetrix Rat Gene 1.0 ST Array 10719303 - -
          Affymetrix Rat Gene 1.0 ST Array 10719300 - -
          Affymetrix Rat Gene 1.0 ST Array 10719301 - -
          Affymetrix Rat Gene 1.0 ST Array 10719302 - -
          Affymetrix Rat Gene 1.0 ST Array 10719297 - -
          Affymetrix Rat Gene 1.0 ST Array 10719299 - -
          Affymetrix Rat Gene 1.1 ST Array 10719294 - -
          Rat Genome 230 2.0 Array 1370873_at - -
          Rat Genome 230 2.0 Array 1368101_at - -
          Rat Genome 230 2.0 Array 1368101_PM_at - -
          Rat Genome 230 2.0 Array 1370873_PM_at - -
          Affymetrix HT Rat Focus Array 1370873_at - -
          Affymetrix HT Rat Focus Array 1368101_at - -
          Affymetrix HT Rat Focus Array 1370873_PM_at - -
          Affymetrix HT Rat Focus Array 1368101_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100038528.rn.1 TC0100004882.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100038529.rn.1 TC0100004882.rn.1 RTA TC
          Rat Expression Set 230 1370873_PM_at - -
          Rat Expression Set 230 1370873_at - -
          Rat Expression Set 230 1368101_PM_at - -
          Rat Expression Set 230 1368101_at - -
          Affymetrix Rat Gene 1.0 ST Array 10719294 - -
          Rat Genome U34 Set X14265_at - -
          Rat Genome U34 Set rc_AA944426_at - -
          Rat Genome U34 Set rc_AI180372_f_at - -
          Rat Genome U34 Set X14265_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21438860 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374160 - -
          RatRef-12 Expression BeadChip ILMN_50866 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline