Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00754976_m1
          See other KNG2 GE Assays ›
          Gene Symbol:
          Kng2
          Gene Name:
          kininogen 2
          Gene Aliases:
          KINKG, KINKH, Kng1, Kngk
          Chromosome Location:
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00754976_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kng2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001102418.1 NP_001095888.1 1248 74
          NM_012741.1 NP_036873.1 9 - 10 1149 74
          GenBank mRNA BC088155.1 - 1248 7369450 74
          L29428.1 - 9 - 10 1149 74

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kng2

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of endopeptidase activity
          cysteine-type endopeptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P296167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2584 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83496 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P296167 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P296167 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P296167 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P296167 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31363 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10940439 - -
          Affymetrix Rat Gene 1.1 ST Array 10755135 - -
          Affymetrix Rat Gene 1.1 ST Array 10940439 - -
          Rat Genome 230 2.0 Array 1371124_a_at - -
          Rat Genome 230 2.0 Array 1369226_at - -
          Rat Genome 230 2.0 Array 1387050_PM_s_at - -
          Rat Genome 230 2.0 Array 1387050_s_at - -
          Rat Genome 230 2.0 Array 1371124_PM_a_at - -
          Rat Genome 230 2.0 Array 1369225_PM_at - -
          Rat Genome 230 2.0 Array 1369226_PM_at - -
          Rat Genome 230 2.0 Array 1369225_at - -
          Affymetrix HT Rat Focus Array 1371124_PM_a_at - -
          Affymetrix HT Rat Focus Array 1371124_a_at - -
          Affymetrix HT Rat Focus Array 1369225_at - -
          Affymetrix HT Rat Focus Array 1369225_PM_at - -
          Rat Expression Set 230 1369225_at - -
          Rat Expression Set 230 1369226_PM_at - -
          Rat Expression Set 230 1369225_PM_at - -
          Rat Expression Set 230 1371124_PM_a_at - -
          Rat Expression Set 230 1387050_PM_s_at - -
          Rat Expression Set 230 1387050_s_at - -
          Rat Expression Set 230 1369226_at - -
          Rat Expression Set 230 1371124_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10940439 - -
          Rat Genome U34 Set M14369exon#2_at - -
          Rat Genome U34 Set K02814_g_at - -
          Rat Genome U34 Set K02814_at - -
          Rat Genome U34 Set rc_AA924618_at - -
          Rat Genome U34 Set K02814_at - -
          Rat Genome U34 Set K02814_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21338317 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline