Search
Search
| Gene Symbol: Cstb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012838.2 | NP_036970.1 | 2 - 3 | 260 | 89 | |
| GenBank mRNA | BC084725.1 | - | 2 - 3 | 248 | 7318993 | 89 |
| CF976177.1 | - | 2 - 3 | 261 | 89 | ||
| FQ220397.1 | - | 2 - 3 | 244 | 89 | ||
| FQ221069.1 | - | 2 - 3 | 220 | 89 | ||
| FQ222613.1 | - | 2 - 3 | 215 | 89 | ||
| X54737.1 | - | 2 - 3 | 252 | 89 | ||
Set Membership: |
|