Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00755009_m1
          See other ATP1B3 GE Assays ›
          Gene Symbol:
          Atp1b3
          Gene Name:
          ATPase, Na+/K+ transporting, beta 3 polypeptide
          Gene Aliases:
          NKAB3S
          Chromosome Location:
          Chr.8: 104190032 - 104221342 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00755009_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atp1b3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012913.1 NP_037045.1 1 - 2 253 91
          GenBank mRNA BC061719.1 - 1 - 2 256 5598284 91
          D84450.1 - 1 - 2 253 91

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atp1b3

          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          potassium ion transport
          sodium ion transport
          cellular sodium ion homeostasis
          potassium ion import
          metal ion transport
          cellular potassium ion homeostasis
          positive regulation of ATPase activity
          sodium ion export from cell
          protein stabilization
          protein localization to plasma membrane
          membrane repolarization
          positive regulation of potassium ion transmembrane transporter activity
          positive regulation of sodium ion export from cell
          positive regulation of potassium ion import
          ATPase activator activity
          sodium:potassium-exchanging ATPase activity
          ATPase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P555125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858961 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176258 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97840 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45208 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101398 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P555125 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P555125 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P555125 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P555125 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700934989 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102732 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57353 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12883 - -
          Agilent Rat cDNA Microarray (G4105A) 700934989 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10919380 - -
          Affymetrix Rat Gene 1.0 ST Array 10910192 - -
          Affymetrix Rat Gene 1.0 ST Array 10919379 - -
          Affymetrix Rat Gene 1.0 ST Array 10919378 - -
          Affymetrix Rat Gene 1.0 ST Array 10919381 - -
          Affymetrix Rat Gene 1.1 ST Array 10910191 - -
          Affymetrix Rat Gene 1.1 ST Array 10919377 - -
          Rat Genome 230 2.0 Array 1398300_PM_at - -
          Rat Genome 230 2.0 Array 1398300_at - -
          Affymetrix HT Rat Focus Array 1398300_at - -
          Affymetrix HT Rat Focus Array 1398300_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800025299.rn.1 TC0800003168.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800025301.rn.1 TC0800003168.rn.1 RTA TC
          Rat Expression Set 230 1398300_at - -
          Rat Expression Set 230 1398300_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10919377 - -
          Rat Genome U34 Set D84450_at - -
          Rat Genome U34 Set rc_AA943304_at - -
          Rat Genome U34 Set rc_AA943304_g_at - -
          Rat Genome U34 Set D84450_at - -
          Rat Genome U34 Set rc_AA943304_g_at - -
          Rat Genome U34 Set D84450_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21044593 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369650 - -
          RatRef-12 Expression BeadChip ILMN_69740 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline