Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00755195_m1
          See other PRDX2 GE Assays ›
          Gene Symbol:
          Prdx2
          Gene Name:
          peroxiredoxin 2
          Gene Aliases:
          Tdpx1
          Chromosome Location:
          Chr.19: 26084816 - 26090095 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00755195_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prdx2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017169.1 NP_058865.1 3 - 4 307 95
          XM_006255244.1 XP_006255306.1 3 - 4 488 95
          GenBank mRNA BC058481.1 - 3 - 4 334 6921522 95
          FQ211079.1 - 3 - 4 323 95
          FQ215547.1 - 3 - 4 359 95
          FQ221561.1 - 3 - 4 328 95
          FQ225602.1 - 3 - 4 330 95
          FQ227282.1 - 3 - 4 330 95
          FQ227580.1 - 3 - 4 321 95
          FQ233937.1 - 3 - 4 330 95
          FQ234358.1 - 3 - 4 359 95
          FQ234566.1 - 3 - 4 425 95
          U06099.1 - 3 - 4 307 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000351.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prdx2

          Panther Classification:

          Molecular Function -

          peroxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          respiratory burst involved in inflammatory response
          response to oxidative stress
          regulation of hydrogen peroxide metabolic process
          removal of superoxide radicals
          positive regulation of blood coagulation
          negative regulation of lipopolysaccharide-mediated signaling pathway
          negative regulation of NF-kappaB transcription factor activity
          response to lipopolysaccharide
          cellular response to oxidative stress
          T cell proliferation
          hydrogen peroxide metabolic process
          hydrogen peroxide catabolic process
          regulation of apoptotic process
          negative regulation of neuron apoptotic process
          negative regulation of T cell differentiation
          thymus development
          homeostasis of number of cells
          oxidation-reduction process
          negative regulation of reactive oxygen species metabolic process
          negative regulation of extrinsic apoptotic signaling pathway
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          molecular_function
          peroxidase activity
          thioredoxin peroxidase activity
          antioxidant activity
          peroxiredoxin activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P14135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178322 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14135 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14135 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P14135 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P14135 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92419 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10810019 - -
          Affymetrix Rat Gene 1.0 ST Array 10708781 - -
          Affymetrix Rat Gene 1.0 ST Array 10810022 - -
          Affymetrix Rat Gene 1.0 ST Array 10810021 - -
          Affymetrix Rat Gene 1.0 ST Array 10810020 - -
          Affymetrix Rat Gene 1.1 ST Array 10810018 - -
          Rat Genome 230 2.0 Array 1367578_PM_at - -
          Rat Genome 230 2.0 Array 1367578_at - -
          Affymetrix HT Rat Focus Array 1367578_PM_at - -
          Affymetrix HT Rat Focus Array 1367578_at - -
          Affymetrix Rat Clariom S Assay PSR1900002746.rn.1 TC1900000351.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900002742.rn.1 TC1900000351.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900002744.rn.1 TC1900000351.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900002743.rn.1 TC1900000351.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900002745.rn.1 TC1900000351.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900002741.rn.1 TC1900000351.rn.1 RTA TC
          Rat Expression Set 230 1367578_PM_at - -
          Rat Expression Set 230 1367578_at - -
          Affymetrix Rat Gene 1.0 ST Array 10810018 - -
          Rat Genome U34 Set U06099_at - -
          Rat Genome U34 Set U06099_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22295125 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364158 - -
          RatRef-12 Expression BeadChip ILMN_48676 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline