Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00755343_m1
          See other CAV3 GE Assays ›
          Gene Symbol:
          Cav3
          Gene Name:
          caveolin 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 144382945 - 144398919 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00755343_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cav3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019155.2 NP_062028.1 1 - 2 245 64
          GenBank mRNA BC084692.1 - 1 - 2 245 7103841 64
          U31968.1 - 1 - 2 243 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001558.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cav3

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          plasma membrane repair
          regulation of heart rate
          negative regulation of protein kinase activity
          triglyceride metabolic process
          endocytosis
          plasma membrane organization
          actin filament organization
          positive regulation of cytosolic calcium ion concentration
          myoblast fusion
          regulation of heart contraction
          protein localization
          positive regulation of cell proliferation
          negative regulation of cardiac muscle hypertrophy
          positive regulation of myotube differentiation
          regulation of skeletal muscle contraction
          myotube differentiation
          cell growth
          regulation of transforming growth factor beta receptor signaling pathway
          cell differentiation
          positive regulation of microtubule polymerization
          cytoplasmic microtubule organization
          membrane raft organization
          detection of muscle stretch
          regulation of signal transduction by receptor internalization
          regulation of membrane potential
          glucose homeostasis
          cholesterol homeostasis
          negative regulation of MAP kinase activity
          negative regulation of MAPK cascade
          negative regulation of cell size
          muscle cell cellular homeostasis
          negative regulation of nitric-oxide synthase activity
          regulation of nerve growth factor receptor activity
          nucleus localization
          regulation of protein kinase B signaling
          regulation of calcium ion transport
          negative regulation of calcium ion transport
          cardiac muscle cell development
          regulation of cardiac muscle contraction
          negative regulation of sarcomere organization
          regulation of ventricular cardiac muscle cell membrane repolarization
          heart trabecula formation
          regulation of ventricular cardiac muscle cell membrane depolarization
          regulation of branching involved in mammary gland duct morphogenesis
          negative regulation of cell growth involved in cardiac muscle cell development
          caveola assembly
          protein localization to plasma membrane
          ventricular cardiac muscle cell action potential
          establishment of protein localization to plasma membrane
          regulation of calcium ion import
          regulation of p38MAPK cascade
          regulation of membrane depolarization during cardiac muscle cell action potential
          negative regulation of membrane depolarization during cardiac muscle cell action potential
          negative regulation of potassium ion transmembrane transporter activity
          regulation of calcium ion transmembrane transporter activity
          negative regulation of potassium ion transmembrane transport
          regulation of sodium ion transmembrane transporter activity
          positive regulation of caveolin-mediated endocytosis
          calcium channel regulator activity
          protein binding
          protein C-terminus binding
          sodium channel regulator activity
          potassium channel inhibitor activity
          enzyme binding
          protein complex binding
          protein complex scaffold
          alpha-tubulin binding
          ion channel binding
          nitric-oxide synthase binding
          connexin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P125539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183939 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113978 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146864 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P125539 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P125539 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P125539 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11954 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P125539 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85558 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95184 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10857665 - -
          Affymetrix Rat Gene 1.0 ST Array 10857666 - -
          Affymetrix Rat Gene 1.0 ST Array 10857664 - -
          Affymetrix Rat Gene 1.1 ST Array 10857663 - -
          Rat Genome 230 2.0 Array 1387814_at - -
          Rat Genome 230 2.0 Array 1387814_PM_at - -
          Affymetrix HT Rat Focus Array 1387814_PM_at - -
          Affymetrix HT Rat Focus Array 1387814_at - -
          Affymetrix Rat Clariom S Assay PSR0400010861.rn.1 TC0400001558.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400010862.rn.1 TC0400001558.rn.1 RTA TC
          Rat Expression Set 230 1387814_PM_at - -
          Rat Expression Set 230 1387814_at - -
          Affymetrix Rat Gene 1.0 ST Array 10857663 - -
          Rat Genome U34 Set rc_AI043968_at - -
          Rat Genome U34 Set rc_AI104707_at - -
          Rat Genome U34 Set rc_AI043968_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21168936 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369419 - -
          RatRef-12 Expression BeadChip ILMN_49947 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline