Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn00755591_m1
See other PDX1 GE Assays ›
Gene Symbol:
Pdx1
Gene Name:
pancreatic and duodenal homeobox 1
Gene Aliases:
Idx1, Ipf1, Stf1
Chromosome Location:
Chr.12: 9496044 - 9501211 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn00755591_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 250 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Inventoried
Catalog #
4331182
Price
284.00
Your Price
Online offer:
266.65
266.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Pdx1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_022852.3
NP_074043.3
1 - 2
508
53
GenBank mRNA
U04833.1
-
1 - 2
508
53
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC1200001050.rn.1
Set Membership:
Ambion siRNA available
Amplicon length less than or equal to 70
3' Most
Recommended Assay
Rattus norvegicus
1700
Inventoried
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene Pdx1
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
liver development
type B pancreatic cell differentiation
glucose metabolic process
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
smoothened signaling pathway
nitric oxide mediated signal transduction
central nervous system development
positive regulation of cell proliferation
negative regulation of cell proliferation
response to wounding
response to glucose
response to iron(II) ion
response to chlorate
organ senescence
positive regulation of cell death
response to organic cyclic compound
morphogenesis of embryonic epithelium
insulin secretion
cell differentiation
pancreas development
exocrine pancreas development
endocrine pancreas development
organ regeneration
response to nutrient levels
positive regulation of insulin secretion
response to vitamin
response to lipid
response to cytokine
response to nicotine
positive regulation of insulin secretion involved in cellular response to glucose stimulus
regulation of cell proliferation
response to drug
glucose homeostasis
response to leucine
response to alkaloid
positive regulation of DNA binding
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
digestive tract development
stem cell differentiation
response to glucocorticoid
detection of glucose
transdifferentiation
response to fatty acid
negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
negative regulation of type B pancreatic cell apoptotic process
RNA polymerase II core promoter proximal region sequence-specific DNA binding
core promoter sequence-specific DNA binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
protein binding
transcription factor binding
protein complex binding
sequence-specific DNA binding
protein heterodimerization activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name
Probe IDs
Targets
Target Type
Agilent Whole Rat Genome Microarray 4x44K v2
A_64_P010623
-
-
Agilent Whole Rat Genome Microarray 4x44K v3
A_64_P010623
-
-
Agilent Whole Rat Genome Microarray 4x44K v3
A_64_P010623
-
-
Agilent SurePrint G3 Rat GE 8x60K Microarray
A_64_P010623
-
-
SurePrint G3 Rat Gene Expression Microarray G4131A
A_44_P491674
-
-
Array Name
Probe IDs
Targets
Target Type
Affymetrix Rat Gene 1.1 ST Array
10899614
-
-
Rat Genome 230 2.0 Array
1375877_PM_at
-
-
Rat Genome 230 2.0 Array
1375877_at
-
-
Rat Genome 230 2.0 Array
1369516_PM_at
-
-
Rat Genome 230 2.0 Array
1369516_at
-
-
Affymetrix HT Rat Focus Array
1369516_at
-
-
Affymetrix HT Rat Focus Array
1369516_PM_at
-
-
Affymetrix Rat Clariom S Assay
PSR1200008857.rn.1
TC1200001050.rn.1
RTA TC
Affymetrix Rat Clariom S Assay
PSR1200008858.rn.1
TC1200001050.rn.1
RTA TC
Rat Expression Set 230
1369516_PM_at
-
-
Rat Expression Set 230
1369516_at
-
-
Rat Genome U34 Set
S67435_s_at
-
-
Array Name
Probe IDs
Targets
Target Type
ABI Rat Genome Survey Microarray
21605836
-
-
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline