Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00755645_m1
          See other ARF2 GE Assays ›
          Gene Symbol:
          Arf2
          Gene Name:
          ADP-ribosylation factor 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 92018562 - 92040370 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00755645_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arf2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024150.2 NP_077064.1 3 - 4 336 64
          XM_006247543.2 XP_006247605.1 3 - 4 434 64
          GenBank mRNA BC098915.1 - 3 - 4 336 7461110 64
          L12381.1 - 2 - 3 383 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000001731.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Arf2

          Panther Classification:

          Molecular Function -

          G-protein

          Gene Ontology Categories:

          Function(s) Process(es)

          small GTPase mediated signal transduction
          protein transport
          vesicle-mediated transport
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P348825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766356 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123021 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92304 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93629 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P348825 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P348825 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P348825 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P348825 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702028401 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702138361 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702425561 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100929 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68120 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11106 - -
          Agilent Rat cDNA Microarray (G4105A) 702138361 - -
          Agilent Rat cDNA Microarray (G4105A) 702425561 - -
          Agilent Rat cDNA Microarray (G4105A) 702028401 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10738718 - -
          Affymetrix Rat Gene 1.0 ST Array 10738722 - -
          Affymetrix Rat Gene 1.0 ST Array 10738721 - -
          Affymetrix Rat Gene 1.0 ST Array 10738719 - -
          Affymetrix Rat Gene 1.0 ST Array 10738717 - -
          Affymetrix Rat Gene 1.0 ST Array 10738720 - -
          Affymetrix Rat Gene 1.0 ST Array 10738723 - -
          Affymetrix Rat Gene 1.1 ST Array 10738716 - -
          Rat Genome 230 2.0 Array 1379014_at - -
          Rat Genome 230 2.0 Array 1368946_PM_at - -
          Rat Genome 230 2.0 Array 1388813_PM_at - -
          Rat Genome 230 2.0 Array 1388813_at - -
          Rat Genome 230 2.0 Array 1368946_at - -
          Rat Genome 230 2.0 Array 1379014_PM_at - -
          Affymetrix HT Rat Focus Array 1368946_at - -
          Affymetrix HT Rat Focus Array 1388813_PM_at - -
          Affymetrix HT Rat Focus Array 1379014_at - -
          Affymetrix HT Rat Focus Array 1379014_PM_at - -
          Affymetrix HT Rat Focus Array 1368946_PM_at - -
          Affymetrix HT Rat Focus Array 1388813_at - -
          Affymetrix Rat Clariom S Assay PSR1000015663.rn.1 TC1000001731.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000015660.rn.1 TC1000001731.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000015661.rn.1 TC1000001731.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000015662.rn.1 TC1000001731.rn.1 RTA TC
          Rat Expression Set 230 1388813_PM_at - -
          Rat Expression Set 230 1388813_at - -
          Rat Expression Set 230 1368946_at - -
          Rat Expression Set 230 1368946_PM_at - -
          Rat Expression Set 230 1379014_at - -
          Rat Expression Set 230 1379014_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10738716 - -
          Rat Genome U34 Set L12381_at - -
          Rat Genome U34 Set rc_AA964530_at - -
          Rat Genome U34 Set rc_AI229753_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20733077 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373540 - -
          RatRef-12 Expression BeadChip ILMN_48990 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline