Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00755673_m1
          See other SLC21A4 GE Assays ›
          Gene Symbol:
          Slc21a4
          Gene Name:
          kidney specific organic anion transporter
          Gene Aliases:
          OAT-K1, OAT-K2, Slco1a3, rOAT-K1, rOAT-K11, rOAT-K13, rOAT-K14, rOAT-K2, rOAT-K3, rOAT-K5, rOAT-K6, rOAT-K7, rOAT-K8, rOAT-K9
          Chromosome Location:
          Chr.4: 176255603 - 176299119 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00755673_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc21a4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030837.1 NP_110464.1 11 - 12 1430 110
          GenBank mRNA AB012662.1 - 10 - 11 1319 110
          AF445993.1 - 9 - 10 1272 110
          AF445994.1 - 9 - 10 1225 110
          AF445995.1 - 9 - 10 1193 110
          AF445996.1 - 9 - 10 1097 110
          AF445997.1 - 10 - 11 1358 110
          AF445998.1 - 8 - 9 1107 110
          AF445999.1 - 7 - 8 974 110
          AF446000.1 - 7 - 8 997 110
          AF446001.1 - 7 - 8 1023 110
          AF446002.1 - 7 - 8 965 110
          AF446003.1 - 6 - 7 832 110
          AF446004.1 - 6 - 7 816 110
          AF446005.1 - 7 - 8 869 110
          AF446006.1 - 8 - 9 1130 110
          BC085730.1 - 4 - 5 512 7110926 110
          D79981.1 - 11 - 12 1430 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400004024.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Transporter 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc21a4

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          organic anion transport
          drug transport
          methotrexate transport
          transporter activity
          organic anion transmembrane transporter activity
          methotrexate transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P123181 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P709191 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P123181 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P709191 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P709191 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P123181 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P123181 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P709191 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P407166 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P709191 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701824179 - -
          Agilent Rat cDNA Microarray (G4105A) 701824179 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10866637 - -
          Affymetrix Rat Gene 1.0 ST Array 10866643 - -
          Affymetrix Rat Gene 1.0 ST Array 10866644 - -
          Affymetrix Rat Gene 1.0 ST Array 10866649 - -
          Affymetrix Rat Gene 1.0 ST Array 10866655 - -
          Affymetrix Rat Gene 1.0 ST Array 10866656 - -
          Affymetrix Rat Gene 1.0 ST Array 10866650 - -
          Affymetrix Rat Gene 1.0 ST Array 10866653 - -
          Affymetrix Rat Gene 1.0 ST Array 10866645 - -
          Affymetrix Rat Gene 1.0 ST Array 10866648 - -
          Affymetrix Rat Gene 1.0 ST Array 10866639 - -
          Affymetrix Rat Gene 1.0 ST Array 10866657 - -
          Affymetrix Rat Gene 1.0 ST Array 10866640 - -
          Affymetrix Rat Gene 1.0 ST Array 10866654 - -
          Affymetrix Rat Gene 1.0 ST Array 10866647 - -
          Affymetrix Rat Gene 1.0 ST Array 10866638 - -
          Affymetrix Rat Gene 1.0 ST Array 10866652 - -
          Affymetrix Rat Gene 1.0 ST Array 10866646 - -
          Affymetrix Rat Gene 1.0 ST Array 10866651 - -
          Affymetrix Rat Gene 1.0 ST Array 10866641 - -
          Affymetrix Rat Gene 1.0 ST Array 10866642 - -
          Affymetrix Rat Gene 1.1 ST Array 10866636 - -
          Rat Genome 230 2.0 Array 1387974_PM_a_at - -
          Rat Genome 230 2.0 Array 1387974_a_at - -
          Rat Genome 230 2.0 Array 1368498_PM_a_at - -
          Rat Genome 230 2.0 Array 1368498_a_at - -
          Affymetrix HT Rat Focus Array 1368498_a_at - -
          Affymetrix HT Rat Focus Array 1387974_a_at - -
          Affymetrix HT Rat Focus Array 1387974_PM_a_at - -
          Affymetrix HT Rat Focus Array 1368498_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR0400028481.rn.1 TC0400004024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400028484.rn.1 TC0400004024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400028478.rn.1 TC0400004024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400028486.rn.1 TC0400004024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400028476.rn.1 TC0400004024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400028490.rn.1 TC0400004024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400028479.rn.1 TC0400004024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400028480.rn.1 TC0400004024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400028482.rn.1 TC0400004024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400028504.rn.1 TC0400004024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400028485.rn.1 TC0400004024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400028491.rn.1 TC0400004024.rn.1 RTA TC
          Rat Expression Set 230 1368498_PM_a_at - -
          Rat Expression Set 230 1387974_PM_a_at - -
          Rat Expression Set 230 1387974_a_at - -
          Rat Expression Set 230 1368498_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10866636 - -
          Rat Genome U34 Set D79981_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20984049 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376681 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0