Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00755832_mH
          See other SERPINA3N GE Assays ›
          Gene Symbol:
          Serpina3n
          Gene Name:
          serine (or cysteine) peptidase inhibitor, clade A, member 3N
          Gene Aliases:
          CPi-26, SPI3, Spi-2.2, Spin2c, Spin3
          Chromosome Location:
          Chr.6: 128073344 - 128080878 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00755832_mH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serpina3n
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031531.1 NP_113719.1 2 - 3 738 74
          GenBank mRNA BC078796.1 - 2 - 3 690 7129287 74
          D00753.1 - 2 - 3 723 74
          FQ209856.1 - 742 74
          HM448398.1 - 634 74
          X16359.1 - 2 - 3 738 74

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Serpina3n

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          inflammatory response
          negative regulation of endopeptidase activity
          response to lipopolysaccharide
          response to vitamin B6
          cellular response to cAMP
          cellular response to interferon-gamma
          cellular response to interleukin-1
          cellular response to interleukin-6
          cellular response to glucocorticoid stimulus
          serine-type endopeptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1000653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966493 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110805 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1000653 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1000653 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1000653 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1000653 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700364565 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16266 - -
          Agilent Rat cDNA Microarray (G4105A) 700364565 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10886624 - -
          Affymetrix Rat Gene 1.0 ST Array 10886622 - -
          Affymetrix Rat Gene 1.0 ST Array 10886626 - -
          Affymetrix Rat Gene 1.0 ST Array 10886623 - -
          Affymetrix Rat Gene 1.0 ST Array 10886625 - -
          Affymetrix Rat Gene 1.1 ST Array 10886621 - -
          Rat Genome 230 2.0 Array 1368224_PM_at - -
          Rat Genome 230 2.0 Array 1368224_at - -
          Affymetrix HT Rat Focus Array 1368224_at - -
          Affymetrix HT Rat Focus Array 1368224_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0600009837.rn.1 TC0600001407.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600009835.rn.1 TC0600001407.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600009834.rn.1 TC0600001407.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600009836.rn.1 TC0600001407.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600009838.rn.1 TC0600001407.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600009839.rn.1 TC0600001407.rn.1 RTA TC
          Rat Expression Set 230 1368224_PM_at - -
          Rat Expression Set 230 1368224_at - -
          Affymetrix Rat Gene 1.0 ST Array 10886621 - -
          Rat Genome U34 Set D00753_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21489031 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372782 - -
          RatRef-12 Expression BeadChip ILMN_60484 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline