Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00755849_m1
          See other CES1E GE Assays ›
          Gene Symbol:
          Ces1e
          Gene Name:
          carboxylesterase 1E
          Gene Aliases:
          Ces1, Es22
          Chromosome Location:
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00755849_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ces1e
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031565.2 NP_113753.2 5 - 6 1001 101
          GenBank mRNA FM062488.1 - 2 - 3 125 101
          FM140223.1 - 3 - 4 319 101
          X81395.1 - 5 - 6 866 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000150.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus Rat Inflammation 1700 Inventoried Rat_Inflammation Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ces1e

          Panther Classification:

          Molecular Function -

          esterase

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          palmitoyl-CoA hydrolase activity
          carboxylic ester hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P115157 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P006498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321298 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9940 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41958 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130365 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98298 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98537 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P115157 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P006498 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P006498 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P115157 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P115157 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P006498 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P498795 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P688604 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65409 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94116 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42349 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27530 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10930690 - -
          Affymetrix Rat Gene 1.0 ST Array 10806211 - -
          Affymetrix Rat Gene 1.0 ST Array 10806212 - -
          Affymetrix Rat Gene 1.0 ST Array 10806214 - -
          Affymetrix Rat Gene 1.0 ST Array 10806217 - -
          Affymetrix Rat Gene 1.0 ST Array 10806219 - -
          Affymetrix Rat Gene 1.0 ST Array 10806213 - -
          Affymetrix Rat Gene 1.0 ST Array 10806216 - -
          Affymetrix Rat Gene 1.0 ST Array 10806220 - -
          Affymetrix Rat Gene 1.0 ST Array 10806218 - -
          Affymetrix Rat Gene 1.0 ST Array 10806215 - -
          Affymetrix Rat Gene 1.1 ST Array 10930689 - -
          Affymetrix Rat Gene 1.1 ST Array 10806210 - -
          Rat Genome 230 2.0 Array 1370352_PM_at - -
          Rat Genome 230 2.0 Array 1370352_at - -
          Rat Genome 230 2.0 Array 1387214_at - -
          Rat Genome 230 2.0 Array 1387214_PM_at - -
          Affymetrix HT Rat Focus Array 1387214_at - -
          Affymetrix HT Rat Focus Array 1387214_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900001228.rn.1 TC1900000150.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900001226.rn.1 TC1900000150.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900001227.rn.1 TC1900000150.rn.1 RTA TC
          Rat Expression Set 230 1387214_at - -
          Rat Expression Set 230 1387214_PM_at - -
          Rat Expression Set 230 1370352_at - -
          Rat Expression Set 230 1370352_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10806210 - -
          Rat Genome U34 Set X81395_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20860815 - -
          ABI Rat Genome Survey Microarray 21755193 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353157 - -
          RatRef-12 Expression BeadChip ILMN_62111 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline