Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00755967_m1
          See other KCNK9 GE Assays ›
          Gene Symbol:
          Kcnk9
          Gene Name:
          potassium channel, two pore domain subfamily K, member 9
          Gene Aliases:
          Task3
          Chromosome Location:
          Chr.7: 113903756 - 113937655 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00755967_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kcnk9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053405.2 NP_445857.2 1 - 2 284 74
          GenBank mRNA AF192366.1 - 1 - 2 374 74
          AF257082.1 - 1 - 2 289 74
          AF391084.1 - 1 - 2 284 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700003306.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Ion Channel 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kcnk9

          Panther Classification:

          Molecular Function -

          ion channel transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of ion transmembrane transport
          cellular potassium ion transport
          potassium ion transmembrane transport
          cochlea development
          voltage-gated ion channel activity
          voltage-gated potassium channel activity
          potassium channel activity
          protein homodimerization activity
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P114934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784765 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8974 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71171 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72225 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132864 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P114934 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P114934 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P114934 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12797 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100099 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11679 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49458 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10904331 - -
          Affymetrix Rat Gene 1.0 ST Array 10904333 - -
          Affymetrix Rat Gene 1.0 ST Array 10904330 - -
          Affymetrix Rat Gene 1.0 ST Array 10904332 - -
          Affymetrix Rat Gene 1.1 ST Array 10904329 - -
          Rat Genome 230 2.0 Array 1369280_at - -
          Rat Genome 230 2.0 Array 1369280_PM_at - -
          Affymetrix HT Rat Focus Array 1369280_PM_at - -
          Affymetrix HT Rat Focus Array 1369280_at - -
          Affymetrix Rat Clariom S Assay PSR0700025243.rn.1 TC0700003306.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700025244.rn.1 TC0700003306.rn.1 RTA TC
          Rat Expression Set 230 1369280_PM_at - -
          Rat Expression Set 230 1369280_at - -
          Affymetrix Rat Gene 1.0 ST Array 10904329 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22023735 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374794 - -
          RatRef-12 Expression BeadChip ILMN_65910 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline