Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00756179_m1
          See other NR2F2 GE Assays ›
          Gene Symbol:
          Nr2f2
          Gene Name:
          nuclear receptor subfamily 2, group F, member 2
          Gene Aliases:
          Tfcoup2
          Chromosome Location:
          Chr.1: 131447671 - 131465745 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00756179_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nr2f2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_080778.2 NP_542956.1 2 - 3 1152 64
          XM_006229366.2 XP_006229428.1 2 - 3 6039 64
          GenBank mRNA AF003944.1 - 2 - 3 1153 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100005746.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Nuclear Receptor 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nr2f2

          Panther Classification:

          Molecular Function -

          C4 zinc finger nuclear receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          in utero embryonic development
          neuron migration
          maternal placenta development
          negative regulation of endothelial cell proliferation
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          skeletal muscle tissue development
          fertilization
          anterior/posterior pattern specification
          radial pattern formation
          negative regulation of endothelial cell migration
          intracellular receptor signaling pathway
          forebrain development
          response to estradiol
          steroid hormone mediated signaling pathway
          negative regulation of cyclin-dependent protein serine/threonine kinase activity
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          blood vessel morphogenesis
          limb development
          placenta blood vessel development
          trophoblast giant cell differentiation
          regulation of transcription involved in lymphatic endothelial cell fate commitment
          retinoic acid binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          steroid hormone receptor activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          zinc ion binding
          protein homodimerization activity
          sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P421333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535678 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 535 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P421333 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P421333 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P421333 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P421333 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101804 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10722737 - -
          Affymetrix Rat Gene 1.0 ST Array 10722738 - -
          Affymetrix Rat Gene 1.0 ST Array 10722739 - -
          Affymetrix Rat Gene 1.1 ST Array 10722736 - -
          Rat Genome 230 2.0 Array 1389554_PM_at - -
          Rat Genome 230 2.0 Array 1389998_PM_at - -
          Rat Genome 230 2.0 Array 1370063_at - -
          Rat Genome 230 2.0 Array 1389998_at - -
          Rat Genome 230 2.0 Array 1389554_at - -
          Rat Genome 230 2.0 Array 1370063_PM_at - -
          Rat Genome 230 2.0 Array 1395863_PM_at - -
          Rat Genome 230 2.0 Array 1395863_at - -
          Affymetrix HT Rat Focus Array 1389998_at - -
          Affymetrix HT Rat Focus Array 1389998_PM_at - -
          Affymetrix HT Rat Focus Array 1370063_at - -
          Affymetrix HT Rat Focus Array 1370063_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100045986.rn.1 TC0100005746.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100045985.rn.1 TC0100005746.rn.1 RTA TC
          Rat Expression Set 230 1370063_at - -
          Rat Expression Set 230 1389998_PM_at - -
          Rat Expression Set 230 1389998_at - -
          Rat Expression Set 230 1370063_PM_at - -
          Rat Expression Set 230 1395863_at - -
          Rat Expression Set 230 1395863_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10722736 - -
          Rat Genome U34 Set rc_AI012183_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22116447 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366431 - -
          RatRef-12 Expression BeadChip ILMN_53018 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline