Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00756233_m1
          See other SLCO1A2 GE Assays ›
          Gene Symbol:
          Slco1a2
          Gene Name:
          solute carrier organic anion transporter family, member 1A2
          Gene Aliases:
          Oatp2, Slc21a5, Slco1a4
          Chromosome Location:
          Chr.4: 175969546 - 176026227 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00756233_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slco1a2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_131906.1 NP_571981.1 14 - 15 1911 134
          XM_008763351.1 XP_008761573.1 14 - 15 1999 134
          GenBank mRNA U88036.1 - 14 - 15 1911 134
          U95011.1 - 15 - 16 2011 134

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400004020.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Transporter 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slco1a2

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          bile acid and bile salt transport
          sodium-independent organic anion transport
          organic anion transmembrane transporter activity
          bile acid transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P003997 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P003997 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P003997 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P003997 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P708480 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P203532 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700503117 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35288 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38314 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9497 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25491 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16075 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37567 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93429 - -
          Agilent Rat cDNA Microarray (G4105A) 700503117 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10867039 - -
          Affymetrix Rat Gene 1.0 ST Array 10867036 - -
          Affymetrix Rat Gene 1.0 ST Array 10867043 - -
          Affymetrix Rat Gene 1.0 ST Array 10867035 - -
          Affymetrix Rat Gene 1.0 ST Array 10867038 - -
          Affymetrix Rat Gene 1.0 ST Array 10867037 - -
          Affymetrix Rat Gene 1.0 ST Array 10867041 - -
          Affymetrix Rat Gene 1.0 ST Array 10867042 - -
          Affymetrix Rat Gene 1.0 ST Array 10867040 - -
          Affymetrix Rat Gene 1.0 ST Array 10867044 - -
          Affymetrix Rat Gene 1.0 ST Array 10867034 - -
          Affymetrix Rat Gene 1.1 ST Array 10867033 - -
          Rat Genome 230 2.0 Array 1387094_at - -
          Rat Genome 230 2.0 Array 1387093_at - -
          Rat Genome 230 2.0 Array 1387093_PM_at - -
          Rat Genome 230 2.0 Array 1387094_PM_at - -
          Affymetrix HT Rat Focus Array 1387093_PM_at - -
          Affymetrix HT Rat Focus Array 1387093_at - -
          Affymetrix HT Rat Focus Array 1387094_at - -
          Affymetrix HT Rat Focus Array 1387094_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400028434.rn.1 TC0400004020.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400028433.rn.1 TC0400004020.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400028436.rn.1 TC0400004020.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400028427.rn.1 TC0400004020.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400028430.rn.1 TC0400004020.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400028432.rn.1 TC0400004020.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400028429.rn.1 TC0400004020.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400028431.rn.1 TC0400004020.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400028435.rn.1 TC0400004020.rn.1 RTA TC
          Rat Expression Set 230 1387093_PM_at - -
          Rat Expression Set 230 1387093_at - -
          Rat Expression Set 230 1387094_PM_at - -
          Rat Expression Set 230 1387094_at - -
          Affymetrix Rat Gene 1.0 ST Array 10867033 - -
          Rat Genome U34 Set U88036_at - -
          Rat Genome U34 Set U88036_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21191806 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline