Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00756257_m1
          See other CBWD1 GE Assays ›
          Gene Symbol:
          Cbwd1
          Gene Name:
          COBW domain containing 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 242819383 - 242861805 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00756257_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cbwd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133535.2 NP_598219.2 3 - 4 342 123
          XM_006231191.2 XP_006231253.1 3 - 4 430 123
          XM_006231192.2 XP_006231254.1 3 - 4 430 123
          XM_006231193.2 XP_006231255.1 3 - 4 430 123
          XM_008760303.1 XP_008758525.1 3 - 4 430 123
          GenBank mRNA AF353305.1 - 3 - 4 398 123
          BC086376.1 - 3 - 4 342 7135348 123

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007456.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cbwd1

          Gene Ontology Categories:

          Process(es)

          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P332472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650549 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121133 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165228 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36010 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16732 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 570 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24749 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62927 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P332472 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P332472 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P332472 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P332472 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52745 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25467 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41335 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54691 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10729493 - -
          Affymetrix Rat Gene 1.0 ST Array 10729494 - -
          Affymetrix Rat Gene 1.0 ST Array 10729496 - -
          Affymetrix Rat Gene 1.0 ST Array 10729499 - -
          Affymetrix Rat Gene 1.0 ST Array 10729490 - -
          Affymetrix Rat Gene 1.0 ST Array 10729495 - -
          Affymetrix Rat Gene 1.0 ST Array 10729489 - -
          Affymetrix Rat Gene 1.0 ST Array 10729498 - -
          Affymetrix Rat Gene 1.0 ST Array 10729488 - -
          Affymetrix Rat Gene 1.0 ST Array 10729492 - -
          Affymetrix Rat Gene 1.0 ST Array 10729497 - -
          Affymetrix Rat Gene 1.0 ST Array 10729501 - -
          Affymetrix Rat Gene 1.0 ST Array 10729500 - -
          Affymetrix Rat Gene 1.0 ST Array 10729491 - -
          Affymetrix Rat Gene 1.0 ST Array 10729487 - -
          Affymetrix Rat Gene 1.1 ST Array 10729485 - -
          Rat Genome 230 2.0 Array 1387514_at - -
          Rat Genome 230 2.0 Array 1380232_at - -
          Rat Genome 230 2.0 Array 1380233_PM_x_at - -
          Rat Genome 230 2.0 Array 1387514_PM_at - -
          Rat Genome 230 2.0 Array 1380233_x_at - -
          Rat Genome 230 2.0 Array 1380232_PM_at - -
          Affymetrix HT Rat Focus Array 1387514_PM_at - -
          Affymetrix HT Rat Focus Array 1387514_at - -
          Affymetrix Rat Clariom S Assay PSR0100060174.rn.1 TC0100007456.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100060173.rn.1 TC0100007456.rn.1 RTA TC
          Rat Expression Set 230 1387514_PM_at - -
          Rat Expression Set 230 1387514_at - -
          Affymetrix Rat Gene 1.0 ST Array 10729485 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21740275 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363173 - -
          RatRef-12 Expression BeadChip ILMN_57333 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline