Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00756303_m1
          See other PRL8A9 GE Assays ›
          Gene Symbol:
          Prl8a9
          Gene Name:
          prolactin family 8, subfamily a, member 9
          Gene Aliases:
          Plpcbeta, Prlpc2
          Chromosome Location:
          Chr.17: 39241133 - 39246941 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00756303_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prl8a9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134385.2 NP_599212.1 4 - 5 390 91
          XM_006253904.1 XP_006253966.1 4 - 5 381 91
          XM_006253905.1 XP_006253967.1 3 - 4 345 91
          GenBank mRNA AF239745.1 - 4 - 5 390 91
          AF239746.1 - 4 - 5 387 91
          AF239747.1 - 4 - 5 390 91
          AF239748.1 - 3 - 4 351 91
          BC100138.1 - 4 - 5 383 7442617 91
          BC127474.1 - 4 - 5 381 7443385 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700000593.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prl8a9

          Panther Classification:

          Molecular Function -

          peptide hormone

          Gene Ontology Categories:

          Process(es)

          hormone activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P058523 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P395625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22692 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14140 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P058523 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P395625 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P058523 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P395625 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P058523 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P395625 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P395625 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66736 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10795031 - -
          Affymetrix Rat Gene 1.0 ST Array 10795032 - -
          Affymetrix Rat Gene 1.0 ST Array 10795033 - -
          Affymetrix Rat Gene 1.0 ST Array 10795030 - -
          Affymetrix Rat Gene 1.0 ST Array 10795035 - -
          Affymetrix Rat Gene 1.0 ST Array 10795034 - -
          Affymetrix Rat Gene 1.0 ST Array 10795029 - -
          Affymetrix Rat Gene 1.0 ST Array 10795028 - -
          Affymetrix Rat Gene 1.1 ST Array 10795027 - -
          Rat Genome 230 2.0 Array 1367841_a_at - -
          Rat Genome 230 2.0 Array 1367841_PM_a_at - -
          Affymetrix HT Rat Focus Array 1367841_a_at - -
          Affymetrix HT Rat Focus Array 1367841_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR1700004085.rn.1 TC1700000593.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700004082.rn.1 TC1700000593.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700004081.rn.1 TC1700000593.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700004088.rn.1 TC1700000593.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700004083.rn.1 TC1700000593.rn.1 RTA TC
          Rat Expression Set 230 1367841_a_at - -
          Rat Expression Set 230 1367841_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10795027 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21239783 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368270 - -
          RatRef-12 Expression BeadChip ILMN_51665 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline