Search
Search
| Gene Symbol: Atp5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139106.1 | NP_620806.1 | 3 - 4 | 439 | 58 | |
| XM_006240845.2 | XP_006240907.1 | 4 - 5 | 954 | 58 | ||
| GenBank mRNA | BC161836.1 | - | 3 - 4 | 2099 | 9098303 | 58 |
| FQ214102.1 | - | 647 | 58 | |||
| FQ223899.1 | - | 3 - 4 | 487 | 58 | ||
| U00926.1 | - | 3 - 4 | 439 | 58 | ||
Set Membership: |
|