Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00756571_m1
          See other CAMK2N1 GE Assays ›
          Gene Symbol:
          Camk2n1
          Gene Name:
          calcium/calmodulin-dependent protein kinase II inhibitor 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 156876706 - 156878486 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00756571_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Camk2n1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173337.1 NP_775459.1 1 - 2 177 113
          GenBank mRNA AF271156.1 - 1 - 2 177 113

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001749.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Camk2n1

          Panther Classification:

          Molecular Function -

          kinase inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of protein kinase activity
          chemical synaptic transmission
          protein kinase inhibitor activity
          calcium-dependent protein kinase inhibitor activity
          protein kinase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P493606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760225 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P493606 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P493606 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P493606 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P493606 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P223630 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700514816 - -
          Agilent Rat cDNA Microarray (G4105A) 700514816 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10873323 - -
          Affymetrix Rat Gene 1.0 ST Array 10873324 - -
          Affymetrix Rat Gene 1.1 ST Array 10873322 - -
          Rat Genome 230 2.0 Array 1370853_at - -
          Rat Genome 230 2.0 Array 1374307_PM_at - -
          Rat Genome 230 2.0 Array 1370853_PM_at - -
          Rat Genome 230 2.0 Array 1374307_at - -
          Rat Genome 230 2.0 Array 1389876_PM_at - -
          Rat Genome 230 2.0 Array 1389876_at - -
          Affymetrix HT Rat Focus Array 1370853_PM_at - -
          Affymetrix HT Rat Focus Array 1370853_at - -
          Affymetrix HT Rat Focus Array 1389876_at - -
          Affymetrix HT Rat Focus Array 1389876_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500013026.rn.1 TC0500001749.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500013027.rn.1 TC0500001749.rn.1 RTA TC
          Rat Expression Set 230 1370853_PM_at - -
          Rat Expression Set 230 1389876_PM_at - -
          Rat Expression Set 230 1389876_at - -
          Rat Expression Set 230 1370853_at - -
          Affymetrix Rat Gene 1.0 ST Array 10873322 - -
          Rat Genome U34 Set rc_AA858621_at - -
          Rat Genome U34 Set rc_AA858621_g_at - -
          Rat Genome U34 Set rc_AA817703_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21343621 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352131 - -
          RatRef-12 Expression BeadChip ILMN_50141 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline