Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00788175_m1
          See other PXK GE Assays ›
          Gene Symbol:
          Pxk
          Gene Name:
          PX domain containing serine/threonine kinase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 18548209 - 18616617 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00788175_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pxk
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_182821.1 NP_877973.1 12 - 13 1256 117
          XM_006251775.1 XP_006251837.1 12 - 13 1284 117
          XM_006251776.2 XP_006251838.1 12 - 13 1284 117
          XM_008770531.1 XP_008768753.1 10 - 11 1774 117
          XM_008770532.1 XP_008768754.1 10 - 11 1774 117
          XM_008770533.1 XP_008768755.1 10 - 11 1774 117
          XM_008770534.1 XP_008768756.1 10 - 11 1774 117
          XM_008770535.1 XP_008768757.1 10 - 11 1774 117
          XM_008770536.1 XP_008768758.1 10 - 11 1774 117
          XM_008770537.1 XP_008768759.1 10 - 11 1774 117
          XM_008770538.1 XP_008768760.1 10 - 11 1774 117
          XM_008770539.1 XP_008768761.1 9 - 10 1283 117
          XM_008770540.1 XP_008768762.1 11 - 12 1149 117
          GenBank mRNA AY327408.1 - 12 - 13 1256 117
          BC098901.1 - 12 - 13 1198 7127537 117

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500001415.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pxk

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          inflammatory response
          biological_process
          negative regulation of ATPase activity
          negative regulation of ion transport
          modulation of synaptic transmission
          molecular_function
          actin binding
          protein kinase activity
          ATP binding
          protein C-terminus binding
          phosphatidylinositol binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P036788 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P100123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512613 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84248 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78103 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15714 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161275 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122915 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168080 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P100123 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P036788 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P036788 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P100123 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P100123 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P036788 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P172941 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70355 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3897 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34807 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45941 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71388 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79352 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10782669 - -
          Affymetrix Rat Gene 1.0 ST Array 10782675 - -
          Affymetrix Rat Gene 1.0 ST Array 10782687 - -
          Affymetrix Rat Gene 1.0 ST Array 10782684 - -
          Affymetrix Rat Gene 1.0 ST Array 10782670 - -
          Affymetrix Rat Gene 1.0 ST Array 10782688 - -
          Affymetrix Rat Gene 1.0 ST Array 10782674 - -
          Affymetrix Rat Gene 1.0 ST Array 10782672 - -
          Affymetrix Rat Gene 1.0 ST Array 10782680 - -
          Affymetrix Rat Gene 1.0 ST Array 10782686 - -
          Affymetrix Rat Gene 1.0 ST Array 10782673 - -
          Affymetrix Rat Gene 1.0 ST Array 10782678 - -
          Affymetrix Rat Gene 1.0 ST Array 10782683 - -
          Affymetrix Rat Gene 1.0 ST Array 10782677 - -
          Affymetrix Rat Gene 1.0 ST Array 10782682 - -
          Affymetrix Rat Gene 1.0 ST Array 10782671 - -
          Affymetrix Rat Gene 1.0 ST Array 10782685 - -
          Affymetrix Rat Gene 1.0 ST Array 10782681 - -
          Affymetrix Rat Gene 1.0 ST Array 10782679 - -
          Affymetrix Rat Gene 1.1 ST Array 10782668 - -
          Rat Genome 230 2.0 Array 1390422_PM_at - -
          Rat Genome 230 2.0 Array 1395279_at - -
          Rat Genome 230 2.0 Array 1395279_PM_at - -
          Rat Genome 230 2.0 Array 1390422_at - -
          Affymetrix HT Rat Focus Array 1395279_at - -
          Affymetrix HT Rat Focus Array 1390422_at - -
          Affymetrix HT Rat Focus Array 1395279_PM_at - -
          Affymetrix HT Rat Focus Array 1390422_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1500008973.rn.1 TC1500001415.rn.1 RTA TC
          Rat Expression Set 230 1390422_PM_at - -
          Rat Expression Set 230 1390422_at - -
          Rat Expression Set 230 1395279_at - -
          Rat Expression Set 230 1395279_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10782668 - -
          Rat Genome U34 Set rc_AI070012_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21622626 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355525 - -
          RatRef-12 Expression BeadChip ILMN_66004 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline