Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00820633_g1
          See other NCL GE Assays ›
          Gene Symbol:
          Ncl
          Gene Name:
          nucleolin
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 93369119 - 93377643 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00820633_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ncl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012749.2 NP_036881.2 6 - 7 1175 86
          GenBank mRNA BC085751.1 - 6 - 7 1175 7109097 86
          FJ817497.1 - 6 - 7 614 86

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ncl

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          endocytosis
          spermatogenesis
          positive regulation of tumor necrosis factor production
          negative regulation of apoptotic process
          positive regulation of transcription from RNA polymerase II promoter
          cellular response to lipopolysaccharide
          liver regeneration
          positive regulation of transcription of nuclear large rRNA transcript from RNA polymerase I promoter
          regulation of rRNA processing
          positive regulation of interleukin-6 secretion
          nucleotide binding
          single-stranded DNA binding
          RNA binding
          receptor binding
          calcium ion binding
          protein binding
          protein C-terminus binding
          selenocysteine insertion sequence binding
          rRNA primary transcript binding
          telomeric DNA binding
          histone binding
          identical protein binding
          laminin binding
          sequence-specific DNA binding
          poly(A) RNA binding
          ErbB-4 class receptor binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P370213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784713 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147286 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P370213 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P370213 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P370213 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P370213 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9576 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10929587 - -
          Affymetrix Rat Gene 1.0 ST Array 10929577 - -
          Affymetrix Rat Gene 1.0 ST Array 10929576 - -
          Affymetrix Rat Gene 1.0 ST Array 10929574 - -
          Affymetrix Rat Gene 1.0 ST Array 10779092 - -
          Affymetrix Rat Gene 1.0 ST Array 10929581 - -
          Affymetrix Rat Gene 1.0 ST Array 10929586 - -
          Affymetrix Rat Gene 1.0 ST Array 10929575 - -
          Affymetrix Rat Gene 1.0 ST Array 10929573 - -
          Affymetrix Rat Gene 1.0 ST Array 10929584 - -
          Affymetrix Rat Gene 1.0 ST Array 10929580 - -
          Affymetrix Rat Gene 1.0 ST Array 10929583 - -
          Affymetrix Rat Gene 1.0 ST Array 10929585 - -
          Affymetrix Rat Gene 1.0 ST Array 10929578 - -
          Affymetrix Rat Gene 1.0 ST Array 10929579 - -
          Affymetrix Rat Gene 1.0 ST Array 10929582 - -
          Affymetrix Rat Gene 1.1 ST Array 10779091 - -
          Affymetrix Rat Gene 1.1 ST Array 10929572 - -
          Rat Genome 230 2.0 Array 1398832_at - -
          Rat Genome 230 2.0 Array 1398832_PM_at - -
          Affymetrix HT Rat Focus Array 1398832_at - -
          Affymetrix HT Rat Focus Array 1398832_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0900019615.rn.1 TC0900002392.rn.1 RTA TC
          Rat Expression Set 230 1398832_PM_at - -
          Rat Expression Set 230 1398832_at - -
          Affymetrix Rat Gene 1.0 ST Array 10929572 - -
          Rat Genome U34 Set M55017exon_s_at - -
          Rat Genome U34 Set M55015cds_s_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360147 - -
          RatRef-12 Expression BeadChip ILMN_48076 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline