Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00820710_g1
          See other UBE2I GE Assays ›
          Gene Symbol:
          Ube2i
          Gene Name:
          ubiquitin-conjugating enzyme E2I
          Gene Aliases:
          UbcE2A
          Chromosome Location:
          Chr.10: 14618612 - 14639285 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00820710_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ube2i
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013050.1 NP_037182.1 7 951 62
          XM_006245918.2 XP_006245980.1 7 953 62
          XM_006245919.2 XP_006245981.1 7 1633 62
          XM_006245920.2 XP_006245982.1 7 1028 62
          XM_008767549.1 XP_008765771.1 7 1085 62
          GenBank mRNA BC086324.1 - 7 951 7305067 62
          BC086592.1 - 7 951 7310403 62
          FQ228426.1 - 7 938 62
          FQ229169.1 - 7 938 62
          U54632.1 - 7 951 62

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ube2i

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          ubiquitin-dependent protein catabolic process
          chromosome segregation
          mitotic nuclear division
          regulation of receptor activity
          protein ubiquitination
          protein sumoylation
          positive regulation of intracellular steroid hormone receptor signaling pathway
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          proteasome-mediated ubiquitin-dependent protein catabolic process
          negative regulation of transcription, DNA-templated
          positive regulation of sequence-specific DNA binding transcription factor activity
          cell division
          positive regulation of SUMO transferase activity
          ubiquitin-protein transferase activity
          protein binding
          ATP binding
          protein C-terminus binding
          transcription factor binding
          ligase activity
          SUMO transferase activity
          enzyme binding
          ubiquitin protein ligase binding
          HLH domain binding
          bHLH transcription factor binding
          poly(A) RNA binding
          ubiquitin protein ligase activity
          SUMO conjugating enzyme activity
          RING-like zinc finger domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P126090 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P116702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894538 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79106 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165985 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P116702 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P116702 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P116702 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11682 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74752 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36753 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10741409 - -
          Affymetrix Rat Gene 1.0 ST Array 10737020 - -
          Affymetrix Rat Gene 1.0 ST Array 10741413 - -
          Affymetrix Rat Gene 1.0 ST Array 10741410 - -
          Affymetrix Rat Gene 1.0 ST Array 10741411 - -
          Affymetrix Rat Gene 1.0 ST Array 10741412 - -
          Affymetrix Rat Gene 1.0 ST Array 10741415 - -
          Affymetrix Rat Gene 1.0 ST Array 10776977 - -
          Affymetrix Rat Gene 1.0 ST Array 10741414 - -
          Affymetrix Rat Gene 1.1 ST Array 10776976 - -
          Affymetrix Rat Gene 1.1 ST Array 10737019 - -
          Affymetrix Rat Gene 1.1 ST Array 10741408 - -
          Rat Genome 230 2.0 Array 1396074_PM_at - -
          Rat Genome 230 2.0 Array 1396074_at - -
          Rat Genome 230 2.0 Array 1388474_at - -
          Rat Genome 230 2.0 Array 1396918_PM_at - -
          Rat Genome 230 2.0 Array 1396918_at - -
          Rat Genome 230 2.0 Array 1388474_PM_at - -
          Rat Genome 230 2.0 Array 1370250_PM_at - -
          Rat Genome 230 2.0 Array 1370250_at - -
          Affymetrix HT Rat Focus Array 1388474_at - -
          Affymetrix HT Rat Focus Array 1370250_at - -
          Affymetrix HT Rat Focus Array 1370250_PM_at - -
          Affymetrix HT Rat Focus Array 1388474_PM_at - -
          Rat Expression Set 230 1370250_PM_at - -
          Rat Expression Set 230 1388474_PM_at - -
          Rat Expression Set 230 1370250_at - -
          Rat Expression Set 230 1388474_at - -
          Rat Expression Set 230 1396074_PM_at - -
          Rat Expression Set 230 1396074_at - -
          Affymetrix Rat Gene 1.0 ST Array 10741408 - -
          Affymetrix Rat Gene 1.0 ST Array 10737019 - -
          Rat Genome U34 Set U54632_g_at - -
          Rat Genome U34 Set U54632_at - -
          Rat Genome U34 Set rc_AI104523_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22211570 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361964 - -
          RatRef-12 Expression BeadChip ILMN_49643 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0