Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00820736_g1
          See other GOT2 GE Assays ›
          Gene Symbol:
          Got2
          Gene Name:
          glutamic-oxaloacetic transaminase 2, mitochondrial
          Gene Aliases:
          ASPATA
          Chromosome Location:
          Chr.19: 9587637 - 9613323 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00820736_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Got2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013177.2 NP_037309.1 6 - 7 810 104
          GenBank mRNA BC061792.1 - 6 - 7 758 5597941 104
          M18467.1 - 6 - 7 781 104

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Got2

          Panther Classification:

          Molecular Function -

          transaminase

          Gene Ontology Categories:

          Function(s) Process(es)

          2-oxoglutarate metabolic process
          oxaloacetate metabolic process
          cellular amino acid metabolic process
          aspartate metabolic process
          aspartate biosynthetic process
          aspartate catabolic process
          glutamate metabolic process
          biosynthetic process
          fatty acid transport
          glutamate catabolic process to aspartate
          glutamate catabolic process to 2-oxoglutarate
          dicarboxylic acid metabolic process
          response to ethanol
          L-kynurenine metabolic process
          L-aspartate:2-oxoglutarate aminotransferase activity
          phospholipid binding
          kynurenine-oxoglutarate transaminase activity
          amino acid binding
          enzyme binding
          pyridoxal phosphate binding
          carboxylic acid binding
          protein homodimerization activity
          poly(A) RNA binding
          L-phenylalanine:2-oxoglutarate aminotransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P243238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916636 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111646 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P243238 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P243238 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P243238 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P243238 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700143160 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89259 - -
          Agilent Rat cDNA Microarray (G4105A) 700143160 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10805798 - -
          Affymetrix Rat Gene 1.0 ST Array 10863419 - -
          Affymetrix Rat Gene 1.0 ST Array 10805797 - -
          Affymetrix Rat Gene 1.0 ST Array 10805800 - -
          Affymetrix Rat Gene 1.0 ST Array 10805799 - -
          Affymetrix Rat Gene 1.0 ST Array 10805795 - -
          Affymetrix Rat Gene 1.0 ST Array 10805796 - -
          Affymetrix Rat Gene 1.0 ST Array 10805801 - -
          Affymetrix Rat Gene 1.1 ST Array 10805794 - -
          Affymetrix Rat Gene 1.1 ST Array 10863418 - -
          Rat Genome 230 2.0 Array 1367819_PM_at - -
          Rat Genome 230 2.0 Array 1389871_PM_at - -
          Rat Genome 230 2.0 Array 1367819_at - -
          Rat Genome 230 2.0 Array 1389871_at - -
          Affymetrix HT Rat Focus Array 1389871_PM_at - -
          Affymetrix HT Rat Focus Array 1367819_at - -
          Affymetrix HT Rat Focus Array 1367819_PM_at - -
          Affymetrix HT Rat Focus Array 1389871_at - -
          Affymetrix Rat Clariom S Assay PSR1900000464.rn.1 TC1900000075.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900000461.rn.1 TC1900000075.rn.1 RTA TC
          Rat Expression Set 230 1389871_PM_at - -
          Rat Expression Set 230 1367819_at - -
          Rat Expression Set 230 1389871_at - -
          Rat Expression Set 230 1367819_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10805794 - -
          Rat Genome U34 Set rc_AA892012_at - -
          Rat Genome U34 Set M18467_at - -
          Rat Genome U34 Set rc_AA892012_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20952512 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356903 - -
          RatRef-12 Expression BeadChip ILMN_69217 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline