Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00820810_g1
          See other RPS3A GE Assays ›
          Gene Symbol:
          Rps3a
          Gene Name:
          ribosomal protein S3a
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 185440477 - 185444846 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00820810_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rps3a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017153.1 NP_058849.1 3 - 4 376 78
          GenBank mRNA BC058483.1 - 3 - 4 381 6887884 78
          FQ209727.1 - 3 - 4 376 78
          FQ211699.1 - 3 - 4 377 78
          FQ211708.1 - 3 - 4 376 78
          FQ217809.1 - 3 - 4 376 78
          FQ219020.1 - 3 - 4 376 78
          FQ219404.1 - 3 - 4 375 78
          FQ219522.1 - 3 - 4 377 78
          FQ219543.1 - 3 - 4 377 78
          FQ219867.1 - 3 - 4 377 78
          FQ220002.1 - 3 - 4 376 78
          FQ220027.1 - 3 - 4 376 78
          FQ220037.1 - 3 - 4 377 78
          FQ220045.1 - 3 - 4 376 78
          FQ220135.1 - 3 - 4 377 78
          FQ220260.1 - 3 - 4 376 78
          FQ220261.1 - 3 - 4 377 78
          FQ220317.1 - 3 - 4 376 78
          FQ220504.1 - 3 - 4 376 78
          FQ220577.1 - 3 - 4 377 78
          FQ221185.1 - 3 - 4 377 78
          FQ221431.1 - 3 - 4 377 78
          FQ221460.1 - 3 - 4 376 78
          FQ221931.1 - 3 - 4 376 78
          FQ222056.1 - 3 - 4 376 78
          FQ222736.1 - 3 - 4 377 78
          FQ222773.1 - 3 - 4 377 78
          FQ222846.1 - 3 - 4 406 78
          FQ222869.1 - 3 - 4 376 78
          FQ222876.1 - 3 - 4 376 78
          FQ223535.1 - 3 - 4 383 78
          FQ223540.1 - 3 - 4 376 78
          FQ225080.1 - 3 - 4 377 78
          FQ225145.1 - 3 - 4 376 78
          FQ225713.1 - 3 - 4 375 78
          FQ227696.1 - 3 - 4 376 78
          FQ227805.1 - 3 - 4 377 78
          FQ228312.1 - 3 - 4 377 78
          FQ228317.1 - 3 - 4 376 78
          FQ228376.1 - 3 - 4 377 78
          FQ228558.1 - 3 - 4 376 78
          FQ229234.1 - 3 - 4 376 78
          FQ229520.1 - 3 - 4 376 78
          FQ229870.1 - 3 - 4 376 78
          FQ229960.1 - 3 - 4 78
          FQ231162.1 - 3 - 4 377 78
          FQ231673.1 - 3 - 4 376 78
          FQ232329.1 - 3 - 4 378 78
          FQ232836.1 - 3 - 4 376 78
          FQ232928.1 - 3 - 4 377 78
          FQ233768.1 - 3 - 4 380 78
          FQ234311.1 - 3 - 4 376 78
          FQ234537.1 - 3 - 4 376 78
          X75161.1 - 3 - 4 376 78

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rps3a

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cytoplasmic translation
          translation
          biological_process
          cell differentiation
          negative regulation of apoptotic process
          molecular_function
          structural constituent of ribosome
          translation initiation factor binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P14247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913239 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62596 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14247 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14247 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P14247 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P14247 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700814133 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30612 - -
          Agilent Rat cDNA Microarray (G4105A) 700814133 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10824084 - -
          Affymetrix Rat Gene 1.0 ST Array 10824085 - -
          Affymetrix Rat Gene 1.0 ST Array 10824083 - -
          Affymetrix Rat Gene 1.0 ST Array 10824086 - -
          Affymetrix Rat Gene 1.1 ST Array 10824082 - -
          Rat Genome 230 2.0 Array 1367606_at - -
          Rat Genome 230 2.0 Array 1367606_PM_at - -
          Affymetrix HT Rat Focus Array 1367606_PM_at - -
          Affymetrix HT Rat Focus Array 1367606_at - -
          Affymetrix Rat Clariom S Assay PSR0200026825.rn.1 TC0200003987.rn.1 RTA TC
          Rat Expression Set 230 1367606_at - -
          Rat Expression Set 230 1367606_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10824082 - -
          Rat Genome U34 Set M84716_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21030878 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354829 - -
          RatRef-12 Expression BeadChip ILMN_58431 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline