Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00820818_g1
          See other GPX4 GE Assays ›
          Gene Symbol:
          Gpx4
          Gene Name:
          glutathione peroxidase 4
          Gene Aliases:
          Gshpx-4, Phgpx, gpx-4, snGpx
          Chromosome Location:
          Chr.7: 12516357 - 12519154 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00820818_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpx4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001039849.2 NP_001034938.1 3 - 4 504 67
          NM_017165.2 NP_058861.3 3 - 4 390 67
          GenBank mRNA AB072798.1 - 3 - 4 504 67
          AF274028.1 - 3 - 4 513 67
          AY570513.1 - 3 - 4 492 67
          CB582508.1 - 3 - 4 414 67
          FQ210610.1 - 3 - 4 298 67
          FQ219811.1 - 3 - 4 67
          FQ221663.1 - 3 - 4 320 67
          FQ221760.1 - 3 - 4 298 67
          FQ221899.1 - 3 - 4 298 67
          FQ221976.1 - 3 - 4 301 67
          FQ222299.1 - 3 - 4 298 67
          FQ227764.1 - 3 - 4 457 67
          FQ230991.1 - 3 - 4 286 67
          FQ231108.1 - 3 - 4 356 67
          FQ231686.1 - 3 - 4 298 67
          FQ233099.1 - 3 - 4 298 67
          L24896.1 - 3 - 4 267 67
          U37427.1 - 3 - 4 384 67
          X82679.1 - 3 - 4 277 67

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gpx4

          Panther Classification:

          Molecular Function -

          peroxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          chromatin organization
          glutathione metabolic process
          response to oxidative stress
          multicellular organism development
          spermatogenesis
          aging
          response to estradiol
          hydrogen peroxide catabolic process
          regulation of inflammatory response
          oxidation-reduction process
          glutathione peroxidase activity
          selenium binding
          glutathione binding
          phospholipid-hydroperoxide glutathione peroxidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P129796 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P112467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449427 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P129796 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P112467 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P129796 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P112467 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P129796 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P112467 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P508341 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700174691 - -
          Agilent Rat cDNA Microarray (G4105A) 700174691 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10900625 - -
          Affymetrix Rat Gene 1.0 ST Array 10900627 - -
          Affymetrix Rat Gene 1.0 ST Array 10900624 - -
          Affymetrix Rat Gene 1.0 ST Array 10900619 - -
          Affymetrix Rat Gene 1.0 ST Array 10900620 - -
          Affymetrix Rat Gene 1.0 ST Array 10900623 - -
          Affymetrix Rat Gene 1.0 ST Array 10900622 - -
          Affymetrix Rat Gene 1.0 ST Array 10900621 - -
          Affymetrix Rat Gene 1.0 ST Array 10900626 - -
          Affymetrix Rat Gene 1.1 ST Array 10900618 - -
          Rat Genome 230 2.0 Array 1386871_at - -
          Rat Genome 230 2.0 Array 1386871_PM_at - -
          Rat Genome 230 2.0 Array 1391529_PM_at - -
          Rat Genome 230 2.0 Array 1391529_at - -
          Affymetrix HT Rat Focus Array 1386871_at - -
          Affymetrix HT Rat Focus Array 1391529_at - -
          Affymetrix HT Rat Focus Array 1391529_PM_at - -
          Affymetrix HT Rat Focus Array 1386871_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700017345.rn.1 TC0700002180.rn.1 RTA TC
          Rat Expression Set 230 1386871_PM_at - -
          Rat Expression Set 230 1386871_at - -
          Rat Expression Set 230 1391529_at - -
          Rat Expression Set 230 1391529_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10900618 - -
          Rat Genome U34 Set L24896_s_at - -
          Rat Genome U34 Set L24896_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22065162 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374479 - -
          RatRef-12 Expression BeadChip ILMN_63096 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline