Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00820837_g1
          See other AHCY GE Assays ›
          Gene Symbol:
          Ahcy
          Gene Name:
          adenosylhomocysteinase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 150587833 - 150603057 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00820837_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ahcy
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017201.1 NP_058897.1 9 - 10 1191 122
          GenBank mRNA M15185.1 - 9 - 10 1191 122

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ahcy

          Panther Classification:

          Molecular Function -

          hydrolase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          chronic inflammatory response to antigenic stimulus
          one-carbon metabolic process
          response to nutrient
          S-adenosylhomocysteine catabolic process
          S-adenosylmethionine cycle
          circadian sleep/wake cycle
          homocysteine biosynthetic process
          adenosylhomocysteinase activity
          copper ion binding
          adenyl nucleotide binding
          identical protein binding
          protein self-association
          NAD binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P15271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280716 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144435 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1482 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15271 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15271 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P15271 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15271 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700530238 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26882 - -
          Agilent Rat cDNA Microarray (G4105A) 700530238 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10850980 - -
          Affymetrix Rat Gene 1.0 ST Array 10850984 - -
          Affymetrix Rat Gene 1.0 ST Array 10850981 - -
          Affymetrix Rat Gene 1.0 ST Array 10850987 - -
          Affymetrix Rat Gene 1.0 ST Array 10850986 - -
          Affymetrix Rat Gene 1.0 ST Array 10850983 - -
          Affymetrix Rat Gene 1.0 ST Array 10850985 - -
          Affymetrix Rat Gene 1.1 ST Array 10850979 - -
          Rat Genome 230 2.0 Array 1367798_at - -
          Rat Genome 230 2.0 Array 1367798_PM_at - -
          Affymetrix HT Rat Focus Array 1367798_PM_at - -
          Affymetrix HT Rat Focus Array 1367798_at - -
          Affymetrix Rat Clariom S Assay PSR0300034766.rn.1 TC0300004542.rn.1 RTA TC
          Rat Expression Set 230 1367798_PM_at - -
          Rat Expression Set 230 1367798_at - -
          Affymetrix Rat Gene 1.0 ST Array 10850979 - -
          Rat Genome U34 Set M15185_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22025627 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357474 - -
          RatRef-12 Expression BeadChip ILMN_69991 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-kvkbn:80/100.66.78.154:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline