Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00820849_g1
          See other EEF2 GE Assays ›
          Gene Symbol:
          Eef2
          Gene Name:
          eukaryotic translation elongation factor 2
          Gene Aliases:
          Ef-2
          Chromosome Location:
          Chr.7: 11401501 - 11406771 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00820849_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eef2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017245.2 NP_058941.1 3 - 4 462 79
          GenBank mRNA BC066661.1 - 3 - 4 462 5623322 79
          Y07504.1 - 3 - 4 448 79

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Eef2

          Panther Classification:

          Molecular Function -

          translation elongation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          hematopoietic progenitor cell differentiation
          translational elongation
          positive regulation of translation
          translation elongation factor activity
          GTPase activity
          GTP binding
          translation activator activity
          protein kinase binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P477785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470555 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P477785 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P477785 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P477785 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P477785 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702141227 - -
          Agilent Rat cDNA Microarray (G4105A) 702141227 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10900316 - -
          Affymetrix Rat Gene 1.0 ST Array 10900313 - -
          Affymetrix Rat Gene 1.0 ST Array 10900308 - -
          Affymetrix Rat Gene 1.0 ST Array 10900309 - -
          Affymetrix Rat Gene 1.0 ST Array 10900312 - -
          Affymetrix Rat Gene 1.0 ST Array 10900311 - -
          Affymetrix Rat Gene 1.0 ST Array 10900317 - -
          Affymetrix Rat Gene 1.0 ST Array 10900315 - -
          Affymetrix Rat Gene 1.0 ST Array 10900310 - -
          Affymetrix Rat Gene 1.0 ST Array 10900314 - -
          Affymetrix Rat Gene 1.0 ST Array 10900307 - -
          Affymetrix Rat Gene 1.1 ST Array 10900306 - -
          Rat Genome 230 2.0 Array 1396549_PM_at - -
          Rat Genome 230 2.0 Array 1370003_PM_at - -
          Rat Genome 230 2.0 Array 1396549_at - -
          Rat Genome 230 2.0 Array 1370003_at - -
          Affymetrix HT Rat Focus Array 1370003_at - -
          Affymetrix HT Rat Focus Array 1396549_at - -
          Affymetrix HT Rat Focus Array 1370003_PM_at - -
          Affymetrix HT Rat Focus Array 1396549_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700016595.rn.1 TC0700002120.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700016598.rn.1 TC0700002120.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700016593.rn.1 TC0700002120.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700016597.rn.1 TC0700002120.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700016596.rn.1 TC0700002120.rn.1 RTA TC
          Rat Expression Set 230 1370003_at - -
          Rat Expression Set 230 1370003_PM_at - -
          Rat Expression Set 230 1396549_PM_at - -
          Rat Expression Set 230 1396549_at - -
          Affymetrix Rat Gene 1.0 ST Array 10900306 - -
          Rat Genome U34 Set rc_AA892801_at - -
          Rat Genome U34 Set rc_AA892801_g_at - -
          Rat Genome U34 Set rc_AA963674_at - -
          Rat Genome U34 Set rc_AI178750_at - -
          Rat Genome U34 Set rc_AA892801_g_at - -
          Rat Genome U34 Set rc_AA963674_at - -
          Rat Genome U34 Set rc_AA963674_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21720397 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375111 - -
          RatRef-12 Expression BeadChip ILMN_57001 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-djsrz:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline