Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00820872_g1
          See other BTG1 GE Assays ›
          Gene Symbol:
          Btg1
          Gene Name:
          B-cell translocation gene 1, anti-proliferative
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 37812831 - 37815088 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00820872_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Btg1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017258.1 NP_058954.1 1 - 2 145 63
          GenBank mRNA L26268.1 - 1 - 2 145 63

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Btg1

          Gene Ontology Categories:

          Function(s) Process(es)

          protein methylation
          response to oxidative stress
          spermatogenesis
          spermatid development
          negative regulation of cell proliferation
          response to peptide hormone
          positive regulation of endothelial cell differentiation
          positive regulation of myoblast differentiation
          positive regulation of angiogenesis
          positive regulation of fibroblast apoptotic process
          protein binding
          enzyme binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P14062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366242 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14062 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14062 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P14062 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P14062 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32248 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10895072 - -
          Affymetrix Rat Gene 1.0 ST Array 10895071 - -
          Affymetrix Rat Gene 1.1 ST Array 10895069 - -
          Rat Genome 230 2.0 Array 1367657_at - -
          Rat Genome 230 2.0 Array 1367657_PM_at - -
          Affymetrix HT Rat Focus Array 1367657_PM_at - -
          Affymetrix HT Rat Focus Array 1367657_at - -
          Affymetrix Rat Clariom S Assay PSR0700004653.rn.1 TC0700000569.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004649.rn.1 TC0700000569.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700004652.rn.1 TC0700000569.rn.1 RTA TC
          Rat Expression Set 230 1367657_at - -
          Rat Expression Set 230 1367657_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10895069 - -
          Rat Genome U34 Set L26268_g_at - -
          Rat Genome U34 Set L26268_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22039472 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356848 - -
          RatRef-12 Expression BeadChip ILMN_62433 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline