Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00820952_g1
          See other BET1 GE Assays ›
          Gene Symbol:
          Bet1
          Gene Name:
          Bet1 golgi vesicular membrane trafficking protein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 29082368 - 29092638 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00820952_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Bet1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019251.2 NP_062124.1 3 - 4 286 110
          GenBank mRNA BC062408.1 - 3 - 4 286 5621977 110
          FQ214230.1 - 3 - 4 340 110
          FQ223263.1 - 3 - 4 361 110
          U42755.1 - 3 - 4 330 110

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Bet1

          Panther Classification:

          Molecular Function -

          SNARE protein

          Gene Ontology Categories:

          Function(s) Process(es)

          ER to Golgi vesicle-mediated transport
          protein transport
          vesicle fusion with Golgi apparatus
          syntaxin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P139763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723486 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40390 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P139763 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P139763 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P139763 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P139763 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84153 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82894 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10860862 - -
          Affymetrix Rat Gene 1.0 ST Array 10860860 - -
          Affymetrix Rat Gene 1.0 ST Array 10860864 - -
          Affymetrix Rat Gene 1.0 ST Array 10860861 - -
          Affymetrix Rat Gene 1.0 ST Array 10860859 - -
          Affymetrix Rat Gene 1.0 ST Array 10860863 - -
          Affymetrix Rat Gene 1.1 ST Array 10860858 - -
          Rat Genome 230 2.0 Array 1368881_PM_at - -
          Rat Genome 230 2.0 Array 1372710_PM_at - -
          Rat Genome 230 2.0 Array 1372710_at - -
          Rat Genome 230 2.0 Array 1368881_at - -
          Affymetrix HT Rat Focus Array 1368881_PM_at - -
          Affymetrix HT Rat Focus Array 1372710_at - -
          Affymetrix HT Rat Focus Array 1372710_PM_at - -
          Affymetrix HT Rat Focus Array 1368881_at - -
          Affymetrix Rat Clariom S Assay PSR0400017099.rn.1 TC0400002392.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400017102.rn.1 TC0400002392.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400017097.rn.1 TC0400002392.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400017098.rn.1 TC0400002392.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400017100.rn.1 TC0400002392.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400017101.rn.1 TC0400002392.rn.1 RTA TC
          Rat Expression Set 230 1368881_PM_at - -
          Rat Expression Set 230 1368881_at - -
          Rat Expression Set 230 1372710_PM_at - -
          Rat Expression Set 230 1372710_at - -
          Affymetrix Rat Gene 1.0 ST Array 10860858 - -
          Rat Genome U34 Set U42755_at - -
          Rat Genome U34 Set rc_AI175027_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22070962 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357381 - -
          RatRef-12 Expression BeadChip ILMN_50947 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline