Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00820984_g1
          See other YWHAG GE Assays ›
          Gene Symbol:
          Ywhag
          Gene Name:
          tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, gamma
          Gene Aliases:
          14-3-3G
          Chromosome Location:
          Chr.12: 23789510 - 23817884 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00820984_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ywhag
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019376.2 NP_062249.1 1 - 2 238 71
          XM_006249184.2 XP_006249246.1 1 - 2 366 71
          GenBank mRNA BC127496.1 - 1 - 2 238 8372306 71
          D17447.1 - 1 - 2 201 71
          S55305.1 - 1 - 2 201 71

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ywhag

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein targeting
          signal transduction
          cellular response to insulin stimulus
          regulation of neuron differentiation
          regulation of synaptic plasticity
          regulation of catalytic activity
          actin binding
          protein kinase C binding
          insulin-like growth factor receptor binding
          protein binding
          protein domain specific binding
          enzyme regulator activity
          receptor tyrosine kinase binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P325791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927523 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154806 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142887 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39558 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P325791 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P325791 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P325791 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P325791 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701288627 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73960 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50641 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 574 - -
          Agilent Rat cDNA Microarray (G4105A) 701288627 - -
          Agilent Rat cDNA Microarray (G4105A) 700248596 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10757603 - -
          Affymetrix Rat Gene 1.0 ST Array 10757601 - -
          Affymetrix Rat Gene 1.0 ST Array 10757605 - -
          Affymetrix Rat Gene 1.0 ST Array 10757604 - -
          Affymetrix Rat Gene 1.0 ST Array 10757600 - -
          Affymetrix Rat Gene 1.0 ST Array 10757602 - -
          Affymetrix Rat Gene 1.1 ST Array 10757599 - -
          Rat Genome 230 2.0 Array 1386866_at - -
          Rat Genome 230 2.0 Array 1386866_PM_at - -
          Affymetrix HT Rat Focus Array 1386866_PM_at - -
          Affymetrix HT Rat Focus Array 1386866_at - -
          Affymetrix Rat Clariom S Assay PSR1200003624.rn.1 TC1200000409.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200003626.rn.1 TC1200000409.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200003628.rn.1 TC1200000409.rn.1 RTA TC
          Rat Expression Set 230 1386866_PM_at - -
          Rat Expression Set 230 1386866_at - -
          Affymetrix Rat Gene 1.0 ST Array 10757599 - -
          Rat Genome U34 Set S55305_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22358265 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365061 - -
          RatRef-12 Expression BeadChip ILMN_68677 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline