Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00820986_g1
          See other ATP5H GE Assays ›
          Gene Symbol:
          Atp5h
          Gene Name:
          ATP synthase, H+ transporting, mitochondrial Fo complex, subunit d
          Gene Aliases:
          Atp5jd, Atpq
          Chromosome Location:
          Chr.10: 103967340 - 103972550 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00820986_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atp5h
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019383.2 NP_062256.1 3 - 4 269 84
          GenBank mRNA BC059139.1 - 3 - 4 273 6921557 84
          BC078846.1 - 2 - 3 221 7133871 84
          D10021.1 - 2 - 3 241 84
          D13120.1 - 3 - 4 262 84
          FQ211219.1 - 3 - 4 280 84
          FQ216885.1 - 3 - 4 269 84
          FQ217843.1 - 3 - 4 280 84
          FQ222479.1 - 3 - 4 262 84

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Transporter 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atp5h

          Panther Classification:

          Molecular Function -

          ATP synthase

          Gene Ontology Categories:

          Function(s) Process(es)

          ATP synthesis coupled proton transport
          ATP metabolic process
          hydrogen ion transmembrane transporter activity
          ATPase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467828 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12048 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12048 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12048 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12048 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700484648 - -
          Agilent Rat cDNA Microarray (G4105A) 700484648 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10748955 - -
          Affymetrix Rat Gene 1.0 ST Array 10882156 - -
          Affymetrix Rat Gene 1.0 ST Array 10791823 - -
          Affymetrix Rat Gene 1.0 ST Array 10748956 - -
          Affymetrix Rat Gene 1.0 ST Array 10882157 - -
          Affymetrix Rat Gene 1.0 ST Array 10748954 - -
          Affymetrix Rat Gene 1.0 ST Array 10748957 - -
          Affymetrix Rat Gene 1.1 ST Array 10791822 - -
          Affymetrix Rat Gene 1.1 ST Array 10748953 - -
          Affymetrix Rat Gene 1.1 ST Array 10882155 - -
          Rat Genome 230 2.0 Array 1367622_at - -
          Rat Genome 230 2.0 Array 1367622_PM_at - -
          Affymetrix HT Rat Focus Array 1367622_PM_at - -
          Affymetrix HT Rat Focus Array 1367622_at - -
          Affymetrix Rat Clariom S Assay PSR1000037279.rn.1 TC1000004112.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000037280.rn.1 TC1000004112.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000037276.rn.1 TC1000004112.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000037278.rn.1 TC1000004112.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000037274.rn.1 TC1000004112.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000037283.rn.1 TC1000004112.rn.1 RTA TC
          Rat Expression Set 230 1367622_PM_at - -
          Rat Expression Set 230 1367622_at - -
          Affymetrix Rat Gene 1.0 ST Array 10882155 - -
          Affymetrix Rat Gene 1.0 ST Array 10791822 - -
          Rat Genome U34 Set D13120_s_at - -
          Rat Genome U34 Set rc_AI008292_at - -
          Rat Genome U34 Set rc_AA925814_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21918388 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362182 - -
          RatRef-12 Expression BeadChip ILMN_68012 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline