Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821006_g1
          See other TMSB10 GE Assays ›
          Gene Symbol:
          Tmsb10
          Gene Name:
          thymosin, beta 10
          Gene Aliases:
          Ptmb10, THYb10
          Chromosome Location:
          Chr.4: 100882216 - 100883294 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821006_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmsb10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021261.2 NP_067084.1 2 - 3 172 152
          XM_006236692.2 XP_006236754.1 2 - 3 223 152
          GenBank mRNA AA924288.1 - 1 - 2 152
          BC126092.1 - 2 - 3 141 152
          FQ221466.1 - 2 - 3 170 152
          FQ223015.1 - 2 - 3 170 152
          FQ224120.1 - 2 - 3 172 152
          FQ224544.1 - 2 - 3 170 152
          FQ229044.1 - 2 - 3 171 152
          M17698.1 - 2 - 3 151 152
          M58404.1 - 1 - 2 246 152
          M58405.1 - 2 - 3 153 152

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmsb10

          Gene Ontology Categories:

          Function(s) Process(es)

          actin filament organization
          spermatid development
          actin cytoskeleton organization
          sequestering of actin monomers
          actin monomer binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1060108 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1060108 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1060108 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1060108 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1060108 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701562834 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702119705 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702164442 - -
          Agilent Rat cDNA Microarray (G4105A) 702164442 - -
          Agilent Rat cDNA Microarray (G4105A) 701562834 - -
          Agilent Rat cDNA Microarray (G4105A) 702119705 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10863283 - -
          Affymetrix Rat Gene 1.0 ST Array 10863284 - -
          Affymetrix Rat Gene 1.1 ST Array 10863282 - -
          Rat Genome 230 2.0 Array 1367655_PM_at - -
          Rat Genome 230 2.0 Array 1367655_at - -
          Affymetrix HT Rat Focus Array 1367655_PM_at - -
          Affymetrix HT Rat Focus Array 1367655_at - -
          Affymetrix Rat Clariom S Assay PSR0400021990.rn.1 TC0400003098.rn.1 RTA TC
          Rat Expression Set 230 1367655_at - -
          Rat Expression Set 230 1367655_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10863282 - -
          Rat Genome U34 Set M58404_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21177388 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360851 - -
          RatRef-12 Expression BeadChip ILMN_64936 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline