Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821025_g1
          See other BCL2L2 GE Assays ›
          Gene Symbol:
          Bcl2l2
          Gene Name:
          Bcl2-like 2
          Gene Aliases:
          BCL-W, BCL-WEL, BCL-WS, Bclw
          Chromosome Location:
          Chr.15: 33543769 - 33549165 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821025_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Bcl2l2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021850.2 NP_068622.1 3 - 4 631 64
          XM_006252016.2 XP_006252078.1 3 - 4 821 64
          XM_008770692.1 XP_008768914.1 2 - 3 1102 64
          XM_008770693.1 XP_008768915.1 2 - 3 1178 64
          XM_008770694.1 XP_008768916.1 3 - 4 1241 64
          GenBank mRNA AF096291.1 - 1 - 2 433 64
          AY185098.1 - 3 - 4 602 64
          AY185099.1 - 3 - 4 336 64
          AY185100.1 - 1 - 2 941 64
          BC074021.1 - 3 - 4 631 7105621 64
          FQ212018.1 - 3 - 4 788 64

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Bcl2l2

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          intrinsic apoptotic signaling pathway in response to DNA damage
          negative regulation of apoptotic process
          Sertoli cell proliferation
          extrinsic apoptotic signaling pathway in absence of ligand
          negative regulation of intrinsic apoptotic signaling pathway
          protein binding
          protein homodimerization activity
          protein heterodimerization activity
          BH domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P550346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782155 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5056 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P550346 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P550346 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P550346 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P550346 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P112788 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22814 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10780244 - -
          Affymetrix Rat Gene 1.0 ST Array 10780247 - -
          Affymetrix Rat Gene 1.0 ST Array 10780242 - -
          Affymetrix Rat Gene 1.0 ST Array 10780248 - -
          Affymetrix Rat Gene 1.0 ST Array 10780241 - -
          Affymetrix Rat Gene 1.0 ST Array 10780240 - -
          Affymetrix Rat Gene 1.0 ST Array 10780245 - -
          Affymetrix Rat Gene 1.0 ST Array 10780243 - -
          Affymetrix Rat Gene 1.1 ST Array 10780239 - -
          Rat Genome 230 2.0 Array 1379481_at - -
          Rat Genome 230 2.0 Array 1369611_at - -
          Rat Genome 230 2.0 Array 1371958_at - -
          Rat Genome 230 2.0 Array 1392480_at - -
          Rat Genome 230 2.0 Array 1369611_PM_at - -
          Rat Genome 230 2.0 Array 1379481_PM_at - -
          Rat Genome 230 2.0 Array 1393707_PM_at - -
          Rat Genome 230 2.0 Array 1392480_PM_at - -
          Rat Genome 230 2.0 Array 1371958_PM_at - -
          Rat Genome 230 2.0 Array 1393707_at - -
          Affymetrix HT Rat Focus Array 1369611_PM_at - -
          Affymetrix HT Rat Focus Array 1369611_at - -
          Affymetrix HT Rat Focus Array 1393707_PM_at - -
          Affymetrix HT Rat Focus Array 1393707_at - -
          Affymetrix Rat Clariom S Assay PSR1500003019.rn.1 TC1500000478.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500003020.rn.1 TC1500000478.rn.1 RTA TC
          Rat Expression Set 230 1369611_at - -
          Rat Expression Set 230 1371958_at - -
          Rat Expression Set 230 1369611_PM_at - -
          Rat Expression Set 230 1371958_PM_at - -
          Rat Expression Set 230 1393707_at - -
          Rat Expression Set 230 1392480_PM_at - -
          Rat Expression Set 230 1393707_PM_at - -
          Rat Expression Set 230 1379481_at - -
          Rat Expression Set 230 1392480_at - -
          Rat Expression Set 230 1379481_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10780239 - -
          Rat Genome U34 Set AF096291_at - -
          Rat Genome U34 Set rc_AA942947_at - -
          Rat Genome U34 Set rc_AI236788_at - -
          Rat Genome U34 Set AF096291_at - -
          Rat Genome U34 Set AF096291_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21184284 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372609 - -
          RatRef-12 Expression BeadChip ILMN_69479 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline