Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821028_g1
          See other NRBF2 GE Assays ›
          Gene Symbol:
          Nrbf2
          Gene Name:
          nuclear receptor binding factor 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.20: 22728147 - 22746477 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821028_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nrbf2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022186.1 NP_071522.1 3 - 4 273 65
          XM_008772913.1 XP_008771135.1 2 - 3 249 65
          GenBank mRNA AB024930.1 - 3 - 4 273 65

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nrbf2

          Panther Classification:

          Molecular Function -

          transcription cofactor

          Gene Ontology Categories:

          Function(s)

          transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of ligand-dependent nuclear receptor transcription coactivator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P135229 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30931 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74785 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12151 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P135229 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P135229 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12151 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P135229 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12151 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P841987 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701655830 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700181484 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22023 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75557 - -
          Agilent Rat cDNA Microarray (G4105A) 701655830 - -
          Agilent Rat cDNA Microarray (G4105A) 700181484 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10829814 - -
          Affymetrix Rat Gene 1.0 ST Array 10771403 - -
          Affymetrix Rat Gene 1.0 ST Array 10824843 - -
          Affymetrix Rat Gene 1.0 ST Array 10829811 - -
          Affymetrix Rat Gene 1.0 ST Array 10829812 - -
          Affymetrix Rat Gene 1.0 ST Array 10829815 - -
          Affymetrix Rat Gene 1.0 ST Array 10829813 - -
          Affymetrix Rat Gene 1.1 ST Array 10771402 - -
          Affymetrix Rat Gene 1.1 ST Array 10824842 - -
          Affymetrix Rat Gene 1.1 ST Array 10829810 - -
          Rat Genome 230 2.0 Array 1387152_at - -
          Rat Genome 230 2.0 Array 1387152_PM_at - -
          Affymetrix HT Rat Focus Array 1387152_at - -
          Affymetrix HT Rat Focus Array 1387152_PM_at - -
          Affymetrix Rat Clariom S Assay PSR2000005205.rn.1 TC2000000580.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000005206.rn.1 TC2000000580.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000005207.rn.1 TC2000000580.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000005208.rn.1 TC2000000580.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000005209.rn.1 TC2000000580.rn.1 RTA TC
          Rat Expression Set 230 1387152_at - -
          Rat Expression Set 230 1387152_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10771402 - -
          Affymetrix Rat Gene 1.0 ST Array 10824842 - -
          Affymetrix Rat Gene 1.0 ST Array 10829810 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21568099 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356300 - -
          RatRef-12 Expression BeadChip ILMN_65988 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline