Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821082_g1
          See other RPL36 GE Assays ›
          Gene Symbol:
          Rpl36
          Gene Name:
          ribosomal protein L36
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 10441228 - 10441981 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821082_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpl36
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022504.1 NP_071949.1 1 - 2 109 72
          GenBank mRNA BC058475.1 - 2 - 3 171 6888196 72
          FQ216992.1 - 1 - 2 111 72
          FQ222487.1 - 2 - 3 130 72
          X68284.1 - 1 - 2 109 72

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rpl36

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          structural constituent of ribosome
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P129629 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P032629 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P129629 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P032629 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P129629 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P032629 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P032629 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P129629 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P789727 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10772983 - -
          Affymetrix Rat Gene 1.0 ST Array 10931411 - -
          Affymetrix Rat Gene 1.0 ST Array 10931412 - -
          Affymetrix Rat Gene 1.0 ST Array 10772985 - -
          Affymetrix Rat Gene 1.0 ST Array 10880684 - -
          Affymetrix Rat Gene 1.0 ST Array 10806015 - -
          Affymetrix Rat Gene 1.0 ST Array 10820503 - -
          Affymetrix Rat Gene 1.0 ST Array 10710847 - -
          Affymetrix Rat Gene 1.0 ST Array 10931413 - -
          Affymetrix Rat Gene 1.0 ST Array 10890859 - -
          Affymetrix Rat Gene 1.0 ST Array 10747911 - -
          Affymetrix Rat Gene 1.0 ST Array 10760456 - -
          Affymetrix Rat Gene 1.1 ST Array 10710846 - -
          Affymetrix Rat Gene 1.1 ST Array 10806014 - -
          Affymetrix Rat Gene 1.1 ST Array 10772982 - -
          Affymetrix Rat Gene 1.1 ST Array 10772984 - -
          Affymetrix Rat Gene 1.1 ST Array 10890858 - -
          Affymetrix Rat Gene 1.1 ST Array 10880683 - -
          Affymetrix Rat Gene 1.1 ST Array 10760455 - -
          Affymetrix Rat Gene 1.1 ST Array 10820502 - -
          Affymetrix Rat Gene 1.1 ST Array 10747910 - -
          Affymetrix Rat Gene 1.1 ST Array 10931410 - -
          Rat Genome 230 2.0 Array 1398301_PM_at - -
          Rat Genome 230 2.0 Array 1398301_at - -
          Affymetrix Rat Clariom S Assay PSR0900012127.rn.1 TC0900001465.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900012126.rn.1 TC0900001465.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900012125.rn.1 TC0900001465.rn.1 RTA TC
          Rat Expression Set 230 1398301_PM_at - -
          Rat Expression Set 230 1398301_at - -
          Affymetrix Rat Gene 1.0 ST Array 10931410 - -
          Affymetrix Rat Gene 1.0 ST Array 10890858 - -
          Affymetrix Rat Gene 1.0 ST Array 10820502 - -
          Affymetrix Rat Gene 1.0 ST Array 10806014 - -
          Affymetrix Rat Gene 1.0 ST Array 10710846 - -
          Affymetrix Rat Gene 1.0 ST Array 10772982 - -
          Affymetrix Rat Gene 1.0 ST Array 10880683 - -
          Affymetrix Rat Gene 1.0 ST Array 10772984 - -
          Affymetrix Rat Gene 1.0 ST Array 10760455 - -
          Affymetrix Rat Gene 1.0 ST Array 10747910 - -
          Rat Genome U34 Set rc_AA892123_at - -
          Rat Genome U34 Set rc_AI009003_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21639446 - -
          Array Name Probe IDs Targets Target Type
          RatRef-12 Expression BeadChip ILMN_65115 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline