Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821140_g1
          See other RAB5A GE Assays ›
          Gene Symbol:
          Rab5a
          Gene Name:
          RAB5A, member RAS oncogene family
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 5910 - 6065 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821140_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rab5a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022692.1 NP_073183.1 440 112
          GenBank mRNA AF072935.1 - 440 112
          BC161848.1 - 766 9025210 112

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rab5a

          Gene Ontology Categories:

          Function(s) Process(es)

          endocytosis
          endosome organization
          small GTPase mediated signal transduction
          positive regulation of smooth muscle cell migration
          protein transport
          vesicle-mediated transport
          synaptic vesicle recycling
          early endosome to late endosome transport
          positive regulation of exocytosis
          regulation of long-term neuronal synaptic plasticity
          positive regulation of smooth muscle cell proliferation
          regulation of endosome size
          regulation of filopodium assembly
          receptor internalization involved in canonical Wnt signaling pathway
          regulation of synaptic vesicle exocytosis
          GTPase activity
          GTP binding
          guanyl nucleotide binding
          GDP binding
          GDP-dissociation inhibitor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P038362 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877050 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145279 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152593 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121163 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63770 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P038362 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12299 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12299 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P038362 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P038362 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12299 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12299 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99582 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52060 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66131 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10926150 - -
          Affymetrix Rat Gene 1.0 ST Array 10926143 - -
          Affymetrix Rat Gene 1.0 ST Array 10926145 - -
          Affymetrix Rat Gene 1.0 ST Array 10926146 - -
          Affymetrix Rat Gene 1.0 ST Array 10926144 - -
          Affymetrix Rat Gene 1.0 ST Array 10773165 - -
          Affymetrix Rat Gene 1.0 ST Array 10926149 - -
          Affymetrix Rat Gene 1.0 ST Array 10926147 - -
          Affymetrix Rat Gene 1.0 ST Array 10926148 - -
          Affymetrix Rat Gene 1.1 ST Array 10773164 - -
          Affymetrix Rat Gene 1.1 ST Array 10926142 - -
          Rat Genome 230 2.0 Array 1387641_at - -
          Rat Genome 230 2.0 Array 1387641_PM_at - -
          Rat Genome 230 2.0 Array 1388800_at - -
          Rat Genome 230 2.0 Array 1388800_PM_at - -
          Affymetrix HT Rat Focus Array 1388800_PM_at - -
          Affymetrix HT Rat Focus Array 1387641_PM_at - -
          Affymetrix HT Rat Focus Array 1388800_at - -
          Affymetrix HT Rat Focus Array 1387641_at - -
          Affymetrix Rat Clariom S Assay PSR1400005018.rn.1 TC1400000732.rn.1 RTA TC
          Rat Expression Set 230 1387641_at - -
          Rat Expression Set 230 1387641_PM_at - -
          Rat Expression Set 230 1388800_PM_at - -
          Rat Expression Set 230 1388800_at - -
          Affymetrix Rat Gene 1.0 ST Array 10926142 - -
          Affymetrix Rat Gene 1.0 ST Array 10773164 - -
          Rat Genome U34 Set AF072935_at - -
          Rat Genome U34 Set rc_AA800305_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21341078 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366994 - -
          RatRef-12 Expression BeadChip ILMN_64550 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline