Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821220_g1
          See other AK2 GE Assays ›
          Gene Symbol:
          Ak2
          Gene Name:
          adenylate kinase 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 147185474 - 147204050 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821220_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ak2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033967.2 NP_001029139.1 1 - 2 162 122
          NM_030986.2 NP_112248.2 1 - 2 162 122
          GenBank mRNA BC061727.1 - 1 - 2 116 5598607 122
          CO570724.1 - 1 - 2 162 122
          D13061.1 - 1 - 2 129 122

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ak2

          Panther Classification:

          Molecular Function -

          nucleotide kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          liver development
          oxidative phosphorylation
          ADP biosynthetic process
          brain development
          AMP metabolic process
          ATP metabolic process
          dATP metabolic process
          adenine metabolic process
          nucleotide phosphorylation
          response to thyroid hormone
          adenylate kinase activity
          ATP binding
          nucleobase-containing compound kinase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P032604 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P032611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310183 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170990 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103752 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P032611 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P032604 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P032604 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P032611 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P032611 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P032604 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P699187 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46872 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61980 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10872263 - -
          Affymetrix Rat Gene 1.0 ST Array 10872265 - -
          Affymetrix Rat Gene 1.0 ST Array 10872261 - -
          Affymetrix Rat Gene 1.0 ST Array 10853796 - -
          Affymetrix Rat Gene 1.0 ST Array 10872264 - -
          Affymetrix Rat Gene 1.0 ST Array 10872262 - -
          Affymetrix Rat Gene 1.1 ST Array 10872260 - -
          Affymetrix Rat Gene 1.1 ST Array 10853795 - -
          Rat Genome 230 2.0 Array 1398349_PM_at - -
          Rat Genome 230 2.0 Array 1386954_PM_at - -
          Rat Genome 230 2.0 Array 1386954_at - -
          Rat Genome 230 2.0 Array 1398349_at - -
          Affymetrix HT Rat Focus Array 1386954_PM_at - -
          Affymetrix HT Rat Focus Array 1398349_PM_at - -
          Affymetrix HT Rat Focus Array 1398349_at - -
          Affymetrix HT Rat Focus Array 1386954_at - -
          Affymetrix Rat Clariom S Assay PSR0500010806.rn.1 TC0500001520.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500010809.rn.1 TC0500001520.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500010808.rn.1 TC0500001520.rn.1 RTA TC
          Rat Expression Set 230 1386954_at - -
          Rat Expression Set 230 1386954_PM_at - -
          Rat Expression Set 230 1398349_PM_at - -
          Rat Expression Set 230 1398349_at - -
          Affymetrix Rat Gene 1.0 ST Array 10853795 - -
          Affymetrix Rat Gene 1.0 ST Array 10872260 - -
          Rat Genome U34 Set rc_AA799466_at - -
          Rat Genome U34 Set rc_AI009194_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21562545 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363976 - -
          RatRef-12 Expression BeadChip ILMN_59955 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline