Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821239_g1
          See other RPL10A GE Assays ›
          Gene Symbol:
          Rpl10a
          Gene Name:
          ribosomal protein L10A
          Gene Aliases:
          -
          Chromosome Location:
          Chr.20: 7908309 - 7910867 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821239_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpl10a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031065.1 NP_112327.1 4 - 5 335 83
          GenBank mRNA BC058468.1 - 4 - 5 342 6919792 83
          FQ221330.1 - 4 - 5 335 83
          FQ221725.1 - 4 - 5 338 83
          FQ222317.1 - 4 - 5 340 83
          X93352.1 - 4 - 5 335 83

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rpl10a

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          RNA binding
          structural constituent of ribosome
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P029516 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P041238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473088 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P041238 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P029516 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P041238 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P029516 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P029516 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P041238 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P140400 - -
          Agilent Rat cDNA Microarray (G4105A) 700174506 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10852741 - -
          Affymetrix Rat Gene 1.0 ST Array 10828739 - -
          Affymetrix Rat Gene 1.0 ST Array 10828742 - -
          Affymetrix Rat Gene 1.0 ST Array 10828743 - -
          Affymetrix Rat Gene 1.0 ST Array 10828741 - -
          Affymetrix Rat Gene 1.0 ST Array 10926966 - -
          Affymetrix Rat Gene 1.0 ST Array 10890300 - -
          Affymetrix Rat Gene 1.0 ST Array 10828740 - -
          Affymetrix Rat Gene 1.0 ST Array 10828738 - -
          Affymetrix Rat Gene 1.1 ST Array 10828737 - -
          Affymetrix Rat Gene 1.1 ST Array 10852740 - -
          Affymetrix Rat Gene 1.1 ST Array 10926965 - -
          Affymetrix Rat Gene 1.1 ST Array 10890299 - -
          Rat Genome 230 2.0 Array 1367580_at - -
          Rat Genome 230 2.0 Array 1367580_PM_at - -
          Affymetrix HT Rat Focus Array 1367580_PM_at - -
          Affymetrix HT Rat Focus Array 1367580_at - -
          Rat Expression Set 230 1367580_at - -
          Rat Expression Set 230 1367580_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10890299 - -
          Affymetrix Rat Gene 1.0 ST Array 10852740 - -
          Affymetrix Rat Gene 1.0 ST Array 10828737 - -
          Affymetrix Rat Gene 1.0 ST Array 10926965 - -
          Rat Genome U34 Set X93352_at - -
          Rat Genome U34 Set R46985_s_at - -
          Rat Genome U34 Set R46985_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21651347 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373792 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371194 - -
          RatRef-12 Expression BeadChip ILMN_51390 - -
          RatRef-12 Expression BeadChip ILMN_57904 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline