Search
Search
| Gene Symbol: Rpl18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031102.2 | NP_112364.1 | 3 - 4 | 208 | 59 | |
| XM_006229161.2 | XP_006229223.1 | 3 - 4 | 689 | 59 | ||
| GenBank mRNA | BC084727.1 | - | 2 - 3 | 200 | 7308070 | 59 |
| CB577548.1 | - | 3 - 4 | 208 | 59 | ||
| DQ403026.1 | - | 2 - 3 | 124 | 59 | ||
| FQ211765.1 | - | 3 - 4 | 226 | 59 | ||
| FQ221110.1 | - | 3 - 4 | 226 | 59 | ||
| FQ222651.1 | - | 3 - 4 | 226 | 59 | ||
| FQ228615.1 | - | 3 - 4 | 229 | 59 | ||
| M20156.1 | - | 2 - 3 | 194 | 59 | ||
Set Membership: |
|