Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821376_g1
          See other TMED2 GE Assays ›
          Gene Symbol:
          Tmed2
          Gene Name:
          transmembrane emp24 domain trafficking protein 2
          Gene Aliases:
          Rnp24
          Chromosome Location:
          Chr.12: 37471259 - 37480346 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821376_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmed2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031722.2 NP_113910.1 1 - 2 228 75
          XM_006249327.2 XP_006249389.1 1 - 2 277 75
          GenBank mRNA BC062036.1 - 1 - 2 228 5623841 75
          X92097.1 - 1 - 2 199 75

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmed2

          Panther Classification:

          Molecular Function -

          vesicle coat protein

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          somitogenesis
          neural tube closure
          embryonic placenta development
          maternal placenta development
          heart looping
          intracellular protein transport
          vesicle targeting
          Golgi organization
          positive regulation of gene expression
          somite rostral/caudal axis specification
          negative regulation of GTPase activity
          multicellular organism growth
          post-anal tail morphogenesis
          embryonic morphogenesis
          protein targeting to plasma membrane
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P121086 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P214054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398119 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87779 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P214054 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P121086 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P121086 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P214054 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P214054 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P121086 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P214054 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12649 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19759 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10761722 - -
          Affymetrix Rat Gene 1.0 ST Array 10761721 - -
          Affymetrix Rat Gene 1.0 ST Array 10761723 - -
          Affymetrix Rat Gene 1.0 ST Array 10761719 - -
          Affymetrix Rat Gene 1.0 ST Array 10761720 - -
          Affymetrix Rat Gene 1.1 ST Array 10761718 - -
          Rat Genome 230 2.0 Array 1389940_at - -
          Rat Genome 230 2.0 Array 1398824_PM_at - -
          Rat Genome 230 2.0 Array 1389940_PM_at - -
          Rat Genome 230 2.0 Array 1398824_at - -
          Affymetrix HT Rat Focus Array 1398824_at - -
          Affymetrix HT Rat Focus Array 1389940_at - -
          Affymetrix HT Rat Focus Array 1389940_PM_at - -
          Affymetrix HT Rat Focus Array 1398824_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200012909.rn.1 TC1200001531.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012911.rn.1 TC1200001531.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012905.rn.1 TC1200001531.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012903.rn.1 TC1200001531.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012907.rn.1 TC1200001531.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012906.rn.1 TC1200001531.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012912.rn.1 TC1200001531.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012902.rn.1 TC1200001531.rn.1 RTA TC
          Rat Expression Set 230 1389940_at - -
          Rat Expression Set 230 1398824_at - -
          Rat Expression Set 230 1389940_PM_at - -
          Rat Expression Set 230 1398824_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10761718 - -
          Rat Genome U34 Set X92097_at - -
          Rat Genome U34 Set rc_AA866460_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22396272 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373896 - -
          RatRef-12 Expression BeadChip ILMN_49565 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline